Skip to Content
Merck
All Photos(1)

Key Documents

MABT1350

Sigma-Aldrich

Anti-Autotaxin (ENPP2) Antibody, clone 7A5

clone 7A5, from mouse

Synonym(s):

Ectonucleotide pyrophosphatase/phosphodiesterase family member 2, EC: 3.1.4.39, E-NPP 2, Extracellular lysophospholipase D, LysoPLD

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€432.00

€154.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00
100 μG
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

7A5, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ENPP2(5168)

General description

Ectonucleotide pyrophosphatase/phosphodiesterase family member 2 (UniProt: Q13822; also known as EC: 3.1.4.39, E-NPP 2, Autotaxin, Extracellular lysophospholipase D, LysoPLD) is encoded by the ENPP2 (also known as ATX, PDNP2) gene (Gene ID: 5168) in human. Autotaxin is a secreted glycoprotein that is present in biological fluids. Its lysophospholipase D catalyzes the production of lysophosphatidic acid (LPA) in extracellular fluids. The level of LPA is shown to be markedly increased in the bronchoalveolar lavage fluid in asthma patients in response to allergens. Lysophosphatidylcholine is shown to be its major substrate. It can also act on sphingosylphosphphorylcholine to produce sphingosine-1-phosphate, a modulator of cell motility. Autotaxin is synthesized with a signal peptide (aa 1-27) and a propeptide (aa 28-35), which is cleaved off by furin in the mature form. Three isoforms of Autotaxin have been reported that are produced by alternative splicing. Isoform 1 is highly expressed in peripheral tissues and isoform 3 is more prominently expressed in the brain. Adipocytes are shown to express only isoform 1. Autotaxin levels are up-regulated in obese subjects with glucose intolerance and during adipogenesis. Autotaxin can also act as an angiogenic factor that stimulates migration of smooth muscle cells and microtubule formation and increased expression of Autotaxin has been reported in various types of cancers. (Ref.: Park, GY., et al. (2013). Am. J. Respir. Crit. Care Med. 188(8); 928 940; Oikonomou, N., et al. (2012). Am. J. Respir. Cell Mol. Biol. 47(5); 566-574).

Specificity

Clone 7A5 detects human and murine autotaxin.

Immunogen

His-tagged recombinant fragment (without the first 28 amino acids) from human Autotaxin.

Application

Anti-Autotaxin (ENPP2), clone 7A5, Cat. No. MABT1350, is a mouse monoclonal antibody that detects Autotaxin (Ectonucleotide pyrophosphatase/phosphodiesterase family member 2) and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50-250 dilution from a representative lot detected Autotaxin (ENPP2) in human kidney and human pancreas tissue sections.

Immunohistochemistry Analysis: A representative lot detected Autotaxin (ENPP2) in Immunohistochemistry applications (Kaffe, E., et. al. (2017). Hepatology. 65(4):1369-1383).

Immunocytochemistry Analysis: A representative lot detected Autotaxin (ENPP2) in Immunocytochemistry applications (Kaffe, E., et. al. (2017). Hepatology. 65(4):1369-1383).

Western Blotting Analysis: A representative lot detected Autotaxin (ENPP2) in Western Blotting applications (Kaffe, E., et. al. (2017). Hepatology. 65(4):1369-1383).

Quality

Evaluated by Western Blotting with recombinant human autotaxin.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected recombinant human autotaxin.

Target description

~100 kDa observed; 98.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service