MIHAWG100
Microfil® funnel with S-Pak®
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white filter, gridded filter
Synonym(s):
Membrane filtration system
About This Item
Recommended Products
material
gridded filter
mixed cellulose esters (MCE)
polypropylene
white filter
Agency
ISO 7704
sterility
sterile; ethylene oxide treated
feature
easily wetted with water
hydrophilic
manufacturer/tradename
Microfil® with S-Pak®
parameter
100 mL sample volume
technique(s)
sterile filtration: suitable
filter diam.
47 mm
matrix
MF-Millipore™
pore size
0.45 μm pore size
input
sample type food(s)
sample type beverage(s)
sample(s) (environmental)
application(s)
bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc
compatibility
for use with EZ-Fit® Filtration Manifold
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Convenient and easy to use
- Sterilized, saves time and expense
- Reduce laboratory workload
- Faster filtration rate
- Consistent recoveries
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service