Skip to Content
Merck

Skip To

MABS45

Anti-Annexin A1 Antibody, clone CPTC-ANXA1-2

clone CPTC-ANXA1-2, from mouse

Synonym(s):

annexin A1, chromobindin-9, annexin I (lipocortin I), Lipocortin I, calpactin-2, Calpactin II, annexin-1, Phospholipase A2 inhibitory protein, Annexin I, Calpactin-2, Annexin-13, Chromobindin-9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 388,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
CPTC-ANXA1-2, monoclonal
Application:
IHC, WB
Citations:
-

€ 388,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1819

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

CPTC-ANXA1-2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANXA1(301)

General description

Annexin A1 (ANXA1) is a calcium and phospholipid binding protein almost entirely restricted to microglia in the CNS. During the peripheral inflammatory response, Annexin A1 is instrumental in the phagocytosis of apoptotic leukocytes. Annexin A1 restores the ability of microglia to differentiate between apoptotic and non-apoptotic cells during inflammatory activation. Current research is geared towards using Annexin A1 as a therapeutic target in neuroinflammation. Annexin A1 has also been shown to be influential in cancer development and proliferation.
~ 39 kDa observed

Immunogen

Full length recombinant protein corresponding to human Annexin A1.

Material obtained from the National Cancer Institute’s Clinical Proteomic Technologies for Cancer (CPTC) program

Application

Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in IH. (http://antibodies.cancer.gov/antibodies/detail/CPTC-ANXA1-2)

Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
This Anti-Annexin A1 Antibody, clone CPTC-ANXA1-2 is validated for use in WB, IH for the detection of Annexin A1.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate
Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Annexin A1 on 10 µg of K562 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABC151HPA011271SAB56000324
Gene Information

human ... ANXA1(301)

Gene Information

human ... ANXA1(301)

Gene Information

human ... ANXA1(301)

Gene Information

human ... ANXA1(301)

antibody form

purified antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

clone

CPTC-ANXA1-2, monoclonal

clone

polyclonal

clone

polyclonal

clone

RM424, monoclonal

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

-

species reactivity

human

species reactivity

human, rat, mouse

species reactivity

mouse, rat, human

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters