Skip to Content
Merck

Skip To

MABS1964

Anti-PEG Antibody, clone AGP4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg
Check Cart for Availability
€ 170,00
100 μg
Check Cart for Availability
€ 458,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
AGP4, monoclonal
Technique(s):
ELISA: suitable, immunohistochemistry: suitable, western blot: suitable
Application:
ELISA, IHC, WB
Citations:
-

€ 170,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

clone

AGP4, monoclonal

purified by

ion-exchange chromatography

species reactivity

mouse, all

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgMκ

General description

Polyethylene glycol-conjugated to E.coli beta-glucuronidase.

Application

Immunohistochemistry Analysis: A representative lot detected PEG in Immunohistochemistry applications (Wang, H., et. al. (2012). J Virol. 86(11):6286-302).

Western Blotting Analysis: A representative lot detected PEG in Western Blotting applications (Ehrlich, G.K., et. al. (2009). J Mol Recognit. 22(2):99-103).

ELISA Analysis: A representative lot detected PEG in ELISA applications (Hao WR et al (2018). Sci. Rep. 8(1); 4256; Chang, C.J., et. al. (2017). Nat Commun. 8(1); 522; Henry, C.E., et. al. (2016). Acta Biomater. 43:61-70).




Biochem/physiol Actions

Clone AGP4 specifically detects proteins conjugated with low to high molecular weight methoxy and amino polyethylene glycols (PEGs).

Physical form

Purified mouse monoclonal antibody IgM in PBS with 0.04% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by ELISA with amino- and methoxy PEG molecules (10K-PEG, 3K-PEG, and 2K-mPEG).

ELISA Analysis: 5 µg/mL of this antibody detected amino- and methoxy-PEG molecules (10K amino-PEG, 3K-amino PEG, and 2K-methoxy PEG).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library