Skip to Content
Merck

Skip To

MABF144D

Anti-CD90.2 (mouse), FITC, clone 30-H12 Antibody

clone 30-H12, from rat, FITC conjugate

Synonym(s):

Thy-1 membrane glycoprotein, Thy-1 antigen, CD90, CD90.2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 97,20

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
30-H12, monoclonal
Application:
FACS
Citations:
-

€ 97,20


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

30-H12, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2b

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Thy1(21838)

General description

The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This rat monoclonal Anti-CD90.2 Antibody (mouse), FITC, clone 30-H12, Cat. No. MABF1449 is validated for use in Flow Cytometry, for the detection of CD90.2.

Physical form

Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2.
Purified by size-exclusion chromatography

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Analysis Note

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected CD90.2 in one million C57Bl/6 splenocytes stained with APC Anti-mouse CD3e.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABF460SAB4700594MABF728
clone

30-H12, monoclonal

clone

30-H12, monoclonal

clone

1aG4/C5, monoclonal

clone

30-H12, monoclonal

conjugate

FITC conjugate

conjugate

PE-Cy7

conjugate

unconjugated

conjugate

PE-Cy7

biological source

rat

biological source

rat

biological source

mouse

biological source

rat

Gene Information

mouse ... Thy1(21838)

Gene Information

mouse ... Thy1(21838)

Gene Information

mouse ... Thy1(21838)

Gene Information

-

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified antibody

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters