Skip to Content
Merck

Skip To

MABE446

Anti-Histone H1° Antibody, clone 34

clone 34, from mouse

Synonym(s):

Histone H1.0, Histone H1(0), Histone H1°

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 481,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
34, monoclonal
Application:
FACS, ICC, IHC, WB
Citations:
6

€ 481,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

34, monoclonal

species reactivity

mouse, bovine, Xenopus, human, rat

species reactivity (predicted by homology)

ox (immunogen homology)

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H1F0(3005)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into 30nm chromatin fibers and higher order structures.
~30 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Immunogen

Recombinant protein corresponding to Ox liver Histone H1°

Application

Anti-Histone H1° Antibody, clone 34 is a highly specific mouse monoclonal antibody, that targets Histone H1 & has been tested in western blotting, ICC, IHC & Flow Cytometry.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blotting Analysis: A representative lot from an independent laboratory detected Histone H1° in Xenopus embryo tissue lysate (Seigneurin, D., et al. (1995). Int J Dev Biol. 39(4):597-603.; Fu, G., et al. (2003). Biol Reprod. 68(5):1569-1576.).

Immunocytochemistry Analyis: A representative lot from an independent laboratory detected Histone H1° in Xenopus unfertilized eggs and early embryos (Fu, G., et al. (2003). Biol Reprod. 68(5):1569-1576.; Adenot, P. G., et al. (2000). J Cell Sci. 113(Pt 16):2897-2907.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Histone H1° in Xenopus embryo tissues (Grunwald, D., et al. (1995). Exp Cell Res. 218(2):586-595.).

Flow Cytometry Analyisis: A representative lot from an independent laboratory detected Histone H1° in FC (Grunwald, D., et al. (1999). Methods Mol Biol. 119:443-454.).

Biochem/physiol Actions

This antibody recognizes Histone H1°.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate
Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Histone H1° in 10 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABE446ABE375MABE71
conjugate

unconjugated

conjugate

-

conjugate

-

conjugate

unconjugated

species reactivity

mouse, bovine, Xenopus, human, rat

species reactivity

human

species reactivity

human, mouse, rat

species reactivity

human, bovine

Gene Information

human ... H1F0(3005)

Gene Information

human ... HIST1H1D(3007)

Gene Information

human ... HIST1H3F(8968)

Gene Information

human ... H2AC21(317772)

clone

34, monoclonal

clone

polyclonal

clone

polyclonal

clone

F152C25W, monoclonal

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
MABE44604053252912412

Questions

Reviews

No rating value

Active Filters