Skip to Content
Merck

Skip To

MABE320

Anti-c-ErbB2/c-Neu (Ab-5) Antibody, clone TA-1

clone TA-1, from mouse

Synonym(s):

Receptor tyrosine-protein kinase erbB-2, Metastatic lymph node gene 19 protein, MLN 19, Proto-oncogene Neu, Proto-oncogene c-ErbB-2, Tyrosine kinase-type cell surface receptor HER2, p185erbB2, CD340

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 416,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
TA-1, monoclonal
Application:
FACS, IF, IP, WB
Citations:
2

€ 416,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1245

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TA-1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable, immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

General description

138 kDa calculated
The ErbB2 (HER2, c-Neu, ErbB2, c-erbB2, or HER2/neu) protein is member of the human epidermal growth factor receptor (HER) family. It is a receptor tyrosine kinase containing an intracellular kinase domain and an extracellular ligand domain, although no endogenous ligand has yet been found. ErbB2 dimerizes with ErbB1, 3, and 4, and can activate different pathways including the PI3K, PLCγ, and MAPK pathways, depending on the ErbB receptor involved. Coupling with ErbB3 activates the PI3K pathway, whereas coupling with any of these three receptors can induce the MAPK pathway. ErbB2 signaling promotes cell proliferation and inhibits apoptosis. ErbB2 is overexpressed in various cancers and may play a role in the development and metastasis of gliomas, and ovarian, breast, lung, and gastric cancer.

Immunogen

Cells overexpressing human c-ErbB2/c-Neu
Epitope: Extracellular domain

Application

Flow Cytometry Analysis: A representative lot from an independent laboratory detected c-ErbB2/c-Neu in HER-2 expressing SK-BR-3 cells (Montgomery, R. B., et al. (2005). Cancer Res. 65(2):650-656.).

Western Blot Analysis: A representative lot from an independent laboratory detected c-ErbB2/c-Neu in HER-2 expressing SK-BR-3 cells (Montgomery, R. B., et al. (2005). Cancer Res. 65(2):650-656.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory precipitated c-ErbB2/c-Neu in IP (McKenzie, S. J., et al. (1989). 4(5):543-548.).

Immunofluorescence Analysis: A representative lot from an independent laboratory detected c-ErbB2/c-Neu in IF (McKenzie, S. J., et al. (1989). 4(5):543-548.).
Research Category
Epigenetics & Nuclear Function

Cell Signaling
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-c-ErbB2/c-Neu (Ab-5) Antibody, clone TA-1 is validated for use in Western Blotting, Flow Cytometry, Immunoprecipitation, Immunofluorescence for the detection of c-ErbB2/c-Neu (Ab-5).

Biochem/physiol Actions

This antibody recognizes the extracellular domain of ErbB2/c-Neu.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ cultured supernatent in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
SK-BR-3 cells
Evaluated by Flow Cytometry in SK-BR-3 cells.

Flow Cytometry Analysis: 0.5 µg of this antibody detected c-ErbB2/c-Neu in 0.5X10E6 SK-BR-3 cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-1127

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABE330SAB1403787PLA0123
Gene Information

human ... ERBB2(2064)

Gene Information

human ... ERBB2(2064)

Gene Information

human ... ERBB2(2064)

Gene Information

rabbit ... ErbB2(2064)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity purified immunoglobulin

clone

TA-1, monoclonal

clone

3B5, monoclonal

clone

3B1, monoclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MABE32004053252597794

Questions

Reviews

No rating value

Active Filters