Skip to Content
Merck

Skip To

MABE162

Anti-IGHMBP2 Antibody, clone11-24

clone mAb11-24, from mouse

Synonym(s):

immunoglobulin mu binding protein 2, DNA-binding protein SMUBP-2, cardiac transcription factor 1, Immunoglobulin mu-binding protein 2, Glial factor 1, ATP-dependent helicase IGHMBP2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 428,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
mAb11-24, monoclonal
Application:
ICC, IP, WB
Citations:
1

€ 428,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mAb11-24, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGHMBP2(3508)

General description

Immunoglobulin mu-binding protein 2 (IGHMBP2) is a nucleic acid binding protein that acts on both RNA and DNA. The mature protein contains an ATP-dependent RNA/DNA helicase domain, an R3H single-stranded nucleic acid-binding domain, and a zinc finger domain. Consistent with these multiple functional domains, in vitro data suggests this protein is involved in and array of cellular functions such as transcriptional activation, DNA replications immunoglobulin class switching, pre-mRNA splicing, and translation.
~110 kDa observed. Band observed at ~60 kDa may be a proteolytic fragment of IGHMBP2. (de Planell-Saguer, M., et al. (2009). Hum Mol Genet. 18(12):2115-2126.)

Immunogen

Recombinant Protein corresponding to human IGHMBP2.

Application

Anti-IGHMBP2 Antibody, clone11-24 is a Mouse Monoclonal Antibody for detection of IGHMBP2 also known as immunoglobulin mu binding protein 2, DNA-binding protein SMUBP-2 & has been validated in WB, ICC & IP.
Immunofluorescence Analysis: A representative lot was used by an independent laboratory in IF. (de Planell-Saguer, M., et al. (2009). Hum Mol Genet. 18(12):2115-2126.)

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in IP. (de Planell-Saguer, M., et al. (2009). Hum Mol Genet. 18(12):2115-2126.)

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blot in Hek293 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected IGHMBP2 on 10 µg of Hek293 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABE232MABS441ABE371
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

serum

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human, mouse, rat

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

clone

mAb11-24, monoclonal

clone

5E10-3E1, monoclonal

clone

4C8H10, monoclonal

clone

polyclonal

Gene Information

human ... IGHMBP2(3508)

Gene Information

human ... IGF2BP2(10644)

Gene Information

human ... WBP2(23558)

Gene Information

human ... PABPN1(8106)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters