Skip to Content
Merck

Skip To

MABE152

Anti-SRp55 Antibody, clone 9-1-56

clone 9-1-56, from mouse

Synonym(s):

splicing factor, arginine/serine-rich 6, arginine/serine-rich splicing factor 6, splicing factor, arginine/serine-rich, 55 kDa, Pre-mRNA-splicing factor SRP55, pre-mRNA splicing factor SRP55

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 428,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
9-1-56, monoclonal
Application:
ICC, IHC, IP, WB
Citations:
6

€ 428,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9-1-56, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRSF6(6431)

General description

Pre-mRNA-splicing factor SRp55 is a member of the serine/arginine-rich (SR) protein family. This group of proteins is found mainly in the nucleus where they play a critical role in regulated and constitutive splicing of precursor mRNAs. SRp55 is involved in the constitutive process for mRNA splicing, and is able to mediate alternative splice site selection. Considered one of the more ubiquitous of the splice factors, SRp55 is upregulated in response to DNA damage when p53 is lacking. SRp55 is significantly phosphorylated on RS domain serine residues.
~ 43 kDa observed. 40 kDa calculated.

Immunogen

Epitope: Not determined
MBP-tagged recombinant protein corresponding to human SRp55.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-SRp55 Antibody, clone 9-1-56 (Mouse Monoclonal Antibody) validated in WB, IP, ICC, IHC to detect SRp55 also known as arginine/serine-rich splicing factor 6, Pre-mRNA-splicing factor SRP55.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl without 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected SRp55 on 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABE126MABE936MABE163
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

Gene Information

human ... SRSF6(6431)

Gene Information

human ... SRSF6(6431)

Gene Information

human ... SRSF1(6426)

Gene Information

human ... SRSF1(6426)

clone

9-1-56, monoclonal

clone

16H3, monoclonal

clone

AK-96, monoclonal

clone

AK96, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters