Skip to Content
Merck
All Photos(3)

Key Documents

MABD167

Sigma-Aldrich

Anti-GATA1 Antibody, clone 4F5

ascites fluid, clone 4F5, from mouse

Synonym(s):

Erythroid transcription factor, Eryf1, GATA-binding factor 1, GATA-1, GF-1, NF-E1 DNA-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€370.00

€370.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4F5, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GATA1(2623)

General description

GATA-1 (GATA1), or alternatively Gata-binding factor 1, GF-1, NF-E1 DNA binding protein, and Erythroid transcription factor (EryF1) encoded by the human gene named GATA1/ERYF1/GF1 is transcriptional activator central for erythroid development. GATA1 also plays critical roles in controlling gene expression in other system as well. Research has shown that GATA1 contributes to the level of alpha synuclein gene expression in Parkinson’s disease and over expression of GATA1 in cortical neurons leads to a decrease in the expression of Rab4b and reduces dendrite branching and maturation, and GATA1 expression is elevated in brains with depressive disorder suggesting that GATA1 may be new target for anti-depressant therapies. EMD-Millipore’s Anti-GATA1 mouse monoclonal antibody has been tested in western blot on K562 cell lysates and paraffin embedded immunohistochemistry on pancreatic cancer tissues and in fluorescent immunocytochemistry on K562 cells in culture.

Immunogen

Purified recombinant fragment of human GATA1 expressed in E. Coli.

Application

Anti-GATA1 Antibody, clone 4F5 is a highly specific mouse monoclonal antibody, that targets GATA & has been tested in western blotting, ICC & IHC.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected GATA1 in K562 cells.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected GATA1 in pancreatic cancer tissue.

Optimal working dilutions must be determined by end user.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected GATA1 in K562 cell lysate.

Target description

~45 kDa observed. Uncharacterized bands may appear in some lysate(s).

Linkage

Replaces: MABE470

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
K562 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanan Zhang et al.
Oncotarget, 7(9), 9859-9875 (2016-02-06)
Angiogenesis is essential for tumor growth. Vascular endothelial growth factor (VEGF) is the most important regulator of tumor angiogenesis. However, how transcription factors interact with histone-modifying enzymes to regulate VEGF transcription and tumor angiogenesis remains unclear. Here, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service