Skip to Content
Merck

Skip To

ABD134

Anti-BMP Antibody15

from rabbit, purified by affinity chromatography

Synonym(s):

Bone morphogenetic protein 15, BMP-15, Growth/differentiation factor 9B, GDF9B, GDF-9B, ODG2, POF4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 273,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-

€ 273,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

immunocytochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BMP15(9210)
mouse ... Bmp15(12155)

General description

Bone morphogenetic protein 15 (BMP15), also known as Growth/differentiation factor 9B (GDF9B), is part of the transforming growth factor-beta (TGF-β) superfamily. BMP15 is an oocyte-specific growth and differentiation factor that stimulates folliculogenesis and development as well as granulosa cell (GC) growth. BMP15 is expressed exclusively in the oocyte. BMP15 defects are linked to ovarian dysgenesis type 2 (ODG2) and premature ovarian failure type 4 (POF4). Potentially, BMP15 could be an oogenesis marker, used to monitor the differentiation of human amniotic fluid stem cells into oocyte-like cells.
~53 kDa observed. Uncharacterized bands may appear in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to human BMP15.

Application

Anti-BMP Antibody15 is a highly specific rabbit polyclonal antibody, that targets BMP & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: 2.5 µg/mL from a representative lot detected BMP15 in NIH/3T3 cells.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate
Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected BMP15 in 15 µg of NIH/3T3 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ZRB1489SAB4503655MABD411
biological source

rabbit

biological source

rabbit (recombinant)

biological source

rabbit

biological source

mouse

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

affinity binding assay: suitable, flow cytometry: suitable, immunocytochemistry: suitable, western blot: suitable

technique(s)

ELISA: 1:40000, western blot: 1:500-1:1000

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, western blot: suitable

species reactivity

mouse

species reactivity

human

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters