Skip to Content
Merck
All Photos(1)

Key Documents

SML3079

Sigma-Aldrich

Ponalrestat

≥98% (HPLC)

Synonym(s):

3-[(4-Bromo-2-fluorophenyl)methyl]-3,4-dihydro-4-oxo-1-phthalazineacetic acid, ICI 128436, MK538

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€83.10
25 MG
€269.00

€83.10


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€83.10
25 MG
€269.00

About This Item

Empirical Formula (Hill Notation):
C17H12BrFN2O3
CAS Number:
Molecular Weight:
391.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€83.10


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(CC(C1=CC=CC=C1C2=O)=NN2CC3=CC=C(C=C3F)Br)=O

InChI

1S/C17H12BrFN2O3/c18-11-6-5-10(14(19)7-11)9-21-17(24)13-4-2-1-3-12(13)15(20-21)8-16(22)23/h1-7H,8-9H2,(H,22,23)

InChI key

LKBFFDOJUKLQNY-UHFFFAOYSA-N

Biochem/physiol Actions

Ponalrestat is an orally available, potent and specific inhibitor of aldehyde reductase 2 (ALR2) that blocks the conversion of glucose to sorbitol. Ponalrestat promotes root elongation through the inhibition of the flavin monooxygenase YUCCA in Arabidopsis.
orally available, potent and specific inhibitor of aldehyde reductase 2 (ALR2)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guang Wang et al.
Chemical research in toxicology, 33(8), 2164-2171 (2020-07-09)
Bile acids (BAs) play essential physiological roles not only by facilitating the absorption and transport of nutrients but also by acting as a complex molecular signaling system. Reduced levels of BAs have been observed in obesity and other metabolic disorders.
Ajmer Singh Grewal et al.
Mini reviews in medicinal chemistry, 16(2), 120-162 (2015-09-10)
Diabetes mellitus occurrence has been associated to the modification of the physiological levels of glucose and is often accompanied by several long-term complications, namely neuropathy, nephropathy, retinopathy, cataract, and cardiovascular. Aldose reductase (AR) is an enzyme of aldoketo reductase super-family
Ying Zhu et al.
The Journal of biological chemistry, 294(52), 19923-19933 (2019-11-17)
Plant development is regulated by both synergistic and antagonistic interactions of different phytohormones, including a complex crosstalk between ethylene and auxin. For instance, auxin and ethylene synergistically control primary root elongation and root hair formation. However, a lack of chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service