Skip to Content
Merck

Skip To

MABC679

Anti-RUNX1 Antibody, clone 2B5

ascites fluid, clone 2B5, from mouse

Synonym(s):

Runt-related transcription factor 1, Acute myeloid leukemia 1 protein, Core-binding factor subunit alpha-2, CBF-alpha-2

Oncogene AML-1, Polyomavirus enhancer-binding protein 2 alpha B subunit, PEA2-alpha B, PEBP2-alpha B, SL3-3 enhancer factor 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 358,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2B5, monoclonal
Application:
WB
Citations:
-

€ 358,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1936

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2B5, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RUNX1(861)

General description

Runt-related transcription factor 1 (RUNX1), also known as Acute myeloid leukemia 1 protein, Core-binding factor subunit alpha-2 (CBF-alpha2), Oncogene AML-1, Polyomavirus enhancer-binding protein 2 alpha B subunit (PEA2-alpha B/PEBP2-alpha B), SL3-3 enhancer factor 1 alpha B subunit, and SL3/AKV core-binding factor alpha B subunit is encoded by the human gene named RUNX1/AML1, CBFA2. RUNX1 is a transcription factor that binds to a wide variety of promoters and enhancers belonging to many types of genes from T cell receptor genes to LCK, IL-3 and GM-CSF. Its major normal role is in the development of blood cells and hematopoiesis. It is expressed in most tissues except in normal brain and heart. Levels are highest in the bone marrow, thymus and peripheral lymphocytes. Defects and or translocations involving RUNX1 are the cause for myeloid leukemias as well as lymphoblastic leukemia. Mutations in RUNX1 function are also linked with many breast cancers. Finally, mutations in RUNX1 have recently been shown to be the cause of cancers in epithelial tissues and linked to squamous cell carcinomas. EMD-Millipore’s Anti-RUNX1 mouse monoclonal antibody has been tested in western blot on Jurkat cell lysates and in ELISA using recombinant RUNX1 protein.
~55 kDa observed. Uncharacterized bands may appear in some lysate(s).

Immunogen

Synthesized peptide of human RUNX1.

Application

Anti-RUNX1 Antibody, clone 2B5 is a highly specific mouse monoclonal antibody & that targets RUNX & has been tested in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Jurkat cell lysate
Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected RUNX1 in Jurkat cell lysate.

Optimal working dilutions must be determined by end user.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0000861M6AV38073SAB4300709
species reactivity

human

species reactivity

mouse, human, rat

species reactivity

mouse, bovine, guinea pig, dog, rabbit, human, rat, horse

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

2B5, monoclonal

clone

2C10, monoclonal

clone

polyclonal

clone

polyclonal

Gene Information

human ... RUNX1(861)

Gene Information

human ... RUNX1(861)

Gene Information

human ... RUNX1(861)

Gene Information

human ... RUNX1(861)

antibody form

ascites fluid

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MABC67904053252917394

Questions

Reviews

No rating value

Active Filters