Skip to Content
Merck

Skip To

MABC653

Anti-S100A4 Antibody, clone 9D5.1

clone 9D5.1, from mouse

Synonym(s):

Protein S100-A4, Calvasculin, Metastasin, Placental calcium-binding protein, Protein Mts1, S100 calcium-binding protein A4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
9D5.1, monoclonal
Application:
ICC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9D5.1, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... S100A4(6275)
mouse ... S100A4(20198)

General description

Protein S100-A4 (UniProt P26447; also known as Calvasculin, Metastasin, Placental calcium-binding protein, Protein Mts1, S100 calcium-binding protein A4) is encoded by the S100A4 (also known as CAPL, MTS1, P9KA, PEL98) gene (Gene ID 6275) in human. S100A4 belongs to a family of low moleclar weight proteins characterized by two calcium-binding helix-loop-helix EF-hand domains (a.a. 12-47 & 50-85 of human S100A4). S100 proteins exist as non-covalent homodimers with cell-specific expression patterns. S100A4 associates with cytoskeletal proteins, such as myosin and F-actin, as well as other actin-binding proteins such as Talin and integrins, and is involved in controlling not just shape and motility of normal cells, but also plays a role in cancer cells invasion/metastasis.
~12 kDa oberved. 11.73/11.72/11.78 kDa (human/mouse/rat) calculated. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: Within EF-hands 1&2.
GST-tagged recombinant human S100A4 internal fragment.

Application

Anti-S100A4 Antibody, clone 9D5.1 is an antibody against S100A4 for use in Western Blotting, Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected S100A4 in HUVECs, HeLa, A431, and NIH/3T3 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Cytoskeletal Signaling

Biochem/physiol Actions

Clone 9D5.1 was raised against human S100-A4 internal fragment that shares high sequence homology with the corresponding region in mouse (96%) and rat (93%) protein.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected S100A4 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABN59MABF854MABF291
antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human, mouse

species reactivity

human

species reactivity

human

species reactivity

human

clone

9D5.1, monoclonal

clone

polyclonal

clone

6B4, monoclonal

clone

5.5, monoclonal

Gene Information

human ... S100A4(6275)
mouse ... S100A4(20198)

Gene Information

rat ... S100B(25742)

Gene Information

human ... S100A9(6280)

Gene Information

human ... S100A8(6279), S100A9(6280)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MABC65304055977350401

Questions

Reviews

No rating value

Active Filters