Skip to Content
Merck

Skip To

MABC40

Anti-LAMP-2 Antibody, clone ABL-93

clone ABL-93, from rat

Synonym(s):

lysosomal-associated membrane protein 2, CD107 antigen-like family member B, CD107b antigen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 467,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
ABL-93, monoclonal
Application:
IF, IP, WB
Citations:
7

€ 467,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ABL-93, monoclonal

species reactivity

mouse

technique(s)

immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lamp2(16784)

General description

Lysosome-associated membrane protein 2 (LAMP-2) is a highly glycosolated protein belonging to the lysosome-associated membrane protein family. This group pf proteins composes a significant portion of the lysosomal membrane and are considered crucial mediators for cellular adhesion by interacting with carbohydrate receptors. LAMP-2 is a profuse and critical component of the lysosomal membrane as it has been implicated in playing a role in lysosomal biogenesis. It also appears to be involved in later stages of phagocytosis and autophagy, where it may act to protect the lysosomal membrane from being digested, assist in signal transduction, moderate levels of acidity within the lysosomal environment, and participate in adhesion. Mutations in LAMP-2 expression have been associated with lysosomal storage diseases or LSDs, along with Danon disease, an X-linked disorder characterized by skeletal myopathy, mental retardation, and hypertrophic cardiomyopathy.
~ 80 kDa observed MW. The calculated MW is 45 kDa LAMP-2 is heavily glycosoated and has been routinely observed at MW′s of 80-120 kDa based on publication data, (Chen, 1985; Ferguson, 2009).

Immunogen

Purified glycoproteins from NIH/3T3 cells, corresponding to mouse LAMP-2.

Application

Anti-LAMP-2 Antibody, clone ABL-93 is an antibody against LAMP-2 for use in WB, IP & IF.
Immunoprecipitation Analysis: This antibody has been shown to immunoprecipitate LAMP-2 in HaNIH cells, (Chen, 1985).

Immunofluorescence Analysis: This antibody has been shown to detect LAMP-2 in mouse brain, (Ferguson, 2009).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody recognizes LAMP-2.

Physical form

Format: Purified
Protein G
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate
Evaluated by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected LAMP-2 in 10 µg of NIH/3T3 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC1766L0668PRS3627
biological source

rat

biological source

mouse

biological source

rabbit

biological source

rabbit

clone

ABL-93, monoclonal

clone

H4B4, monoclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse

species reactivity

mouse, rat, human, monkey

species reactivity

human (predicted), mouse, rat

species reactivity

mouse, human

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Gene Information

mouse ... Lamp2(16784)

Gene Information

human ... LAMP2(3920)

Gene Information

human ... LAMP2(3920)
mouse ... Lamp2(16784)
rat ... Lamp2(24944)

Gene Information

mouse ... Lamp2(16784)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Imene Jaadane et al.
Journal of cellular and molecular medicine, 21(12), 3453-3466 (2017-07-01)
Ageing and alteration of the functions of the retinal pigment epithelium (RPE) are at the origin of lost of vision seen in age-related macular degeneration (AMD). The RPE is known to be vulnerable to high-energy blue light. The white light-emitting
Debashis Dutta et al.
Cell reports, 40(2), 111058-111058 (2022-07-14)
This study underlines the importance of treadmill exercise in reducing α-synuclein (α-syn) spreading in the A53T brain and protecting nigral dopaminergic neurons. Preformed α-syn fibril (PFF) seeding in the internal capsule of young A53T α-syn mice leads to increased spreading
Celia A McKee et al.
Scientific reports, 12(1), 1796-1796 (2022-02-04)
An emerging link between circadian clock function and neurodegeneration has indicated a critical role for the molecular clock in brain health. We previously reported that deletion of the core circadian clock gene Bmal1 abrogates clock function and induces cell-autonomous astrocyte activation. Regulation of



Global Trade Item Number

SKUGTIN
MABC4004053252523854

Questions

Reviews

No rating value

Active Filters