Skip to Content
Merck

Skip To

MABC153

Anti-CKAP4 Antibody, clone 9C4.4

clone 9C4.4, from mouse

Synonym(s):

Cytoskeleton-associated protein 4, 63-kDa cytoskeleton-linking membrane protein, Climp-63, p63

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 378,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
9C4.4, monoclonal
Application:
WB
Citations:
-

€ 378,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9C4.4, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CKAP4(10970)

General description

Cytoskeleton-associated protein 4, also known as 63-kDa cytoskeleton-linking membrane protein or Climp-63 or p63, and encoded by the gene CKAP4, is an integral membrane protein that links the endoplasmic reticulum (ER) to microtubules. The binding of CKAP4 is dependent upon its phosphorylation state: dephosphorylated CKAP4 binds microtubules and phosphorylated CKAP4 does not bind microtubules. CKAP4 is localized to the endoplasmic reticulum membrane and plasma membrane; CKAP4 is also found in the cytoplasm and perinuclear regions upon APF stimulation. CKAP4 is expressed is a wide variety of cell types and tissues and CKAP4 appears to be critical for antibiotic cytotoxicity for certain drugs like Gentamycin which bind the drug and cause CKAP4 to dimerize and malfunction.
~ 65 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: coiled coil domain
GST-tagged recombinant protein corresponding to the coiled coil domain of human CKAP4.

Application

Detect Cytoskeleton-associated protein 4 using this mouse monoclonal antibody, Anti-CKAP4 Antibody & clone 9C4.4 validated for use in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected CKAP4 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA000278ABT10MABN356
biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... CKAP4(10970)

Gene Information

human ... CKAP4(10970)

Gene Information

human ... CKAP5(9793)

Gene Information

human ... FKBP4(2288)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

clone

9C4.4, monoclonal

clone

polyclonal

clone

polyclonal

clone

2H9.1, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters