Skip to Content
Merck

Skip To

MABC150

Anti-LDH-A Antibody, clone 5D2.1

clone 5D2.1, from mouse

Synonym(s):

L-lactate dehydrogenase A chain, LDH-A, Cell proliferation-inducing gene 19 protein, LDH muscle subunit, LDH-M, Renal carcinoma antigen NY-REN-59

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 340,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
5D2.1, monoclonal
Application:
IHC, WB
Citations:
2

€ 340,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB2726

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D2.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LDHA(3939)

General description

L-lactate dehydrogenase A chain (LDHA), also known as Cell proliferation-inducing gene 19 protein, LDH muscle subunit, LDH-M, Renal carcinoma antigen NY-REN-59, and encoded by the gene name LDHA/ PIG19, is a member of the LDH/MDH superfamily and LDH family. Mutations in LDHA have been assoicated with Glycogen storage disease 11 (GSD11), which is a metabolic disorder that results in exertional myoglobinuria, pain, cramps and easy fatigue.
~37 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human LDH-A.

Application

Anti-LDH-A, clone 5D2.1 is an antibody against LDH-A for use in WB, IH.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected LDH-A in human breast cancer sections and human kidney tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected LDH-A in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2108638SAB5700695SAB1402835
biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

mouse

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

species reactivity

human

species reactivity

human

species reactivity

human, mouse, rat

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

5D2.1, monoclonal

clone

polyclonal

clone

polyclonal

clone

2C8, monoclonal

Gene Information

human ... LDHA(3939)

Gene Information

human ... LDHA(3939)

Gene Information

human ... LDHA(3939)

Gene Information

human ... LDHC(3948)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters