Skip to Content
Merck

Skip To

MABC128

Anti-FIP200 Antibody, clone 14E11.2

clone 14E11.2, from mouse

Synonym(s):

RB1-inducible coiled-coil protein, 1FAK family kinase-interacting protein of 200 kDa, FIP200

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 467,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
14E11.2, monoclonal
Application:
ICC, WB
Citations:
4

€ 467,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

14E11.2, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RB1CC1(9821)

General description

Focal adhesion kinase family interacting protein of 200 kDa (FIP200) is a novel protein inhibitor for focal adhesion kinase (FAK). FIP200 inhibits kinase activity and associated cellular functions, such as cell adhesion, spreading, and motility in fibroblasts by binding directly to FAK.

The previously assigned protein identifier Q86YR4 has been merged into Q8TDY2. Full details can be found on the UniProt database.
~200 kDa observed. An uncharacterized band may be observed at ~50 kDa in some cell lysates.

Immunogen

Epitope: Interaction domain
GST-tagged recombinant protein corresponding to the interaction domain of human FIP200.

Application

Anti-FIP200 Antibody, clone 14E11.2 is an antibody against FIP200 for use in Western Blotting, ICC.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blot Analysis: 1 µg/mL from a representative lot detected FIP200 in serum starved and non-serum starved HeLa cell lysate.

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected FIP200 in NIH/3T3, A431, and HeLa cells.

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected FIP200 in untreated and chloroquinone-treated HUVEC cells.

Biochem/physiol Actions

This antibody binds to the interaction domain of FIP200, which is needed for the formation of the ULK-FIP200 complex under starved, autophagy conditions.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected FIP200 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV50467MABE325MABS1754
clone

14E11.2, monoclonal

clone

polyclonal

clone

EA10, monoclonal

clone

2F11, monoclonal

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

mouse

Gene Information

human ... RB1CC1(9821)

Gene Information

human ... RB1CC1(9821)

Gene Information

human ... CDKN1A(1026)

Gene Information

mouse ... Mapk14(26416)

species reactivity

human, mouse

species reactivity

bovine, dog, human, rat, mouse, guinea pig, horse

species reactivity

human

species reactivity

human, mouse, rat

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters