Skip to Content
Merck

Skip To

IM57

Anti-MT1-MMP (Ab-4) Mouse mAb (113-5B7)

liquid, clone 113-5B7, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 384,00

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
113-5B7, monoclonal
Species reactivity:
human, rabbit, rat, mouse
Application:
Citations:
3

€ 384,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

113-5B7, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, rabbit, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (10 µg/mL)
Immunofluorescence
Immunoprecipitation
Paraffin Sections )

isotype

IgG3

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

General description

Purified mouse monoclonal antibody. Recognizes the ~60 kDa MTI-MMP protein in overexpressing cells.
Recognizes the ~60 kDa MTI-MMP protein in HT-1080 cells and in ConA-treated MD-MB-231 cells.
This Anti-MT1-MMP (Ab-4) Mouse mAb (113-5B7) is validated for use in Immunoblotting, Immunofluorescence, Immunoprecipitation, Paraffin Sections for the detection of MT1-MMP (Ab-4).

Immunogen

Human
a synthetic peptide (CDGNFDTVAMLRGEM) corresponding to amino acids 319-333 in the hemopexin-like domain of human MT1-MMP

Application


Immunoblotting (10 g/ml)
Immunofluorescence (see application references)
Immunoprecipitation (see application references)
Paraffin Sections (see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 100 mM sodium phosphate buffer, 0.1% BSA, pH 7.0.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HT-1080 or ConA-treated MD-MB-231 cells

Other Notes

Sakakibara, M., et al. 1999. Cancer85, 231-239.
Mattei, M.G., et al. 1997. Genomics:40, 168.
Okada, A., et al. 1997. J. Cell. Biol.147, 67.
Okumura, Y., et al. 1997. FEBS Lett.402, 181.
Ueno, H., et al. 1997. Cancer Res.57, 2055.
Strongin, A.Y., et al. 1995. J. Biol. Chem.270 5331.
Takino, T., et al. 1995. J. Biol. Chem.270, 23013.
Cottam, D. W. and Rees, R. C., 1993. Intl J. Oncol.2, 861.
Stetler-Stevenson, W. G., et al. 1993. FASEB J.7, 1434.
Fridman, R., et al. 1992. J. Biol. Chem.267, 15389.
Woessner, J. F., 1991. FASEB J.5, 2145.
Liotta, L. A. and Stetler-Stevenson, W. G., 1990. in Sem. Cancer Biol., ed. M. M. Gottesman. Vol. 1(2), 99.
Goldberg, G.I., et al. 1989. PNAS86, 8207.
This antibody cross-reacts slightly with human MMP-3. To induce MT1-MMP in MD-MB-231 cells, treat with 1-5 µg/ml ConA for 24-48 h. Antibody should be titrated for optimal results in individual systems.

Legal Information

Manufactured by Daiichi Fine Chemicals C0., Ltd., Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
IM40OP143IM33
clone

113-5B7, monoclonal

clone

141-7B2, monoclonal

clone

4B11, monoclonal

clone

42-5D11, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

human, rabbit, rat, mouse

species reactivity

human

species reactivity

mouse, human

species reactivity

bovine, rat, mouse, human

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

Gene Information

human ... MMP14(4323)

Gene Information

human ... MMP7(4316)

Gene Information

human ... MDM2(4193)
mouse ... Mdm2(17246)

Gene Information

human ... MMP2(4313)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Stephanie W Watts et al.
American journal of physiology. Heart and circulatory physiology, 292(5), H2438-H2448 (2007-01-24)
Arterial remodeling occurs in response to mechanical and neurohumoral stimuli. We hypothesized that veins, which are not exposed to higher pressures in hypertension, would demonstrate less active remodeling than arteries. We assessed remodeling with two standard measures of arterial remodeling:
Yong-Feng Yang et al.
Scientific reports, 6, 30505-30505 (2016-07-29)
The trabecular meshwork (TM) tissue controls drainage of aqueous humor from the anterior chamber of the eye primarily by regulating extracellular matrix (ECM) remodeling by matrix metalloproteinases (MMPs). Glaucomatous TM tissue is stiffer than age-matched controls, which may be due
Ricardo Cruz-Acuña et al.
Journal of cell science, 132(20) (2019-09-29)
Synthetic hydrogels with controlled physicochemical matrix properties serve as powerful in vitro tools to dissect cell-extracellular matrix (ECM) interactions that regulate epithelial morphogenesis in 3D microenvironments. In addition, these fully defined matrices overcome the lot-to-lot variability of naturally derived materials



Global Trade Item Number

SKUGTIN
IM57-100UG04055977220476

Questions

Reviews

No rating value

Active Filters