Skip to Content
Merck
All Photos(1)

Key Documents

GCR2

Millipore

mPAGE® Gel Caster

2 pack, device for hand casting mini gels, does not include plates or combs

Synonym(s):

gel casting frame

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105305
NACRES:
NC.01
Pricing and availability is not currently available.

packaging

pack of 2

manufacturer/tradename

Millipore

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

W × H

14.8 cm × 5.6 cm , caster dimensions

gel size

8.3 cm × 10.1 cm

compatibility

for use with mPAGE®

  • TurboMix Bis-Tris Gel Casting Kit
  • Combs
  • Mini Spacer Plates
  • Short Plates
  • Mini Gel Tank
  • Mini Wet Transfer System

General description

The mPAGE® Gel Caster offers a leak-free solution for handcasting up to 2 mini protein gels. Gels cast
are 8 cm x 10 cm with a gel thickness of 1.0 mm, 0.75 mm, or 1.5 mm, along
with 10 or 15-well options for sample loading. The mPAGE® Gel Caster has been
designed to simplify hand casting gels and to eliminate leakage during the casting
process.

Components

The gel caster is available as a standalone device and includes
  • mPAGE® Gel Caster Base (2)
  • mPAGE® Gel Caster Frame (2)
  • mPAGE® Gel Caster Sealing Gaskets (2)
  • mPAGE® Gel Releasers (5)

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Protocols

Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.

Questions

  1. Can I use BioRad Mini-PROTEAN Short plates and Spacer plates together with mPAGE Gel caster (GCR2)? Thank you for the answer.

    1 answer
    1. Yes, the Bio-Rad plates are the same size as the MilliporeSigma gel caster plates.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service