Skip to Content
Merck

Skip To

EZHPP

Human Pancreatic Polypeptide (PP) Space Saver (Bulk) ELISA Kit

This Human Pancreatic Polypeptide (PP) ELISA Kit is used to measure & quantify Pancreatic Polypeptide levels in Metabolism & Endocrine research.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
96 wells
Check Cart for Availability
€ 6.370,00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.32
eCl@ss:
32161000
Species reactivity:
human

€ 6.370,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

species reactivity

human

technique(s)

ELISA: suitable

assay range

sensitivity: 12.3 pg/mL
(with a 50 μL sample size), inter-assay cv: 4.4-9.8%
intra-assay cv: 3.3-5.0%
(Serum)

detection method

colorimetric, fluorometric

General description

This Human Pancreatic Polypeptide (PP) ELISA kit is used for the non-radioactive quantification of Human PP in human serum, plasma, tissue extract and cell culture samples. One kit is sufficient to measure 39 unknown samples in duplicate

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders
Room temperature, 3.5 hour assay, 50 μL sample size (serum or plasma)
Used to detect/quantify: Pancreatic Polypeptide

Biochem/physiol Actions

Cross Reactivty
The antibody pair used in this assay is specific to Human PP and has no significant cross-reactivity with NPY, PYY and other gut hormones. This assay does not recognize Rat PP

Other Notes

Please contact Technical Service for linearity of dilution.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EZHNPYRAB0725RAB0064
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

assay range

sensitivity: 12.3 pg/mL
(with a 50 μL sample size), inter-assay cv: 4.4-9.8%
intra-assay cv: 3.3-5.0%
(Serum)

assay range

-

assay range

-

assay range

-

Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

-

detection method

colorimetric, fluorometric

detection method

colorimetric (450nm/590nm)

detection method

colorimetric

detection method

colorimetric

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
EZHPP-40BK04053252686405

Questions

Reviews

No rating value

Active Filters