Skip to Content
Merck

Skip To

DP02A

Anti-SV40 T Antigen (Ab-2) Mouse mAb (PAb416) Agarose Conjugate

slurry (Liquid), clone PAb416, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
0.5 mL
Check Cart for Availability
€ 596,00

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
PAb416, monoclonal
Species reactivity:
SV40-infected cells
Application:
Citations:
-

€ 596,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PAb416, monoclonal

form

slurry (Liquid)

contains

≤0.1% sodium azide as preservative

species reactivity

SV40-infected cells

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

dilution

(Immunoprecipitation (15 µL suspension/sample)
Affinity Purification )

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells (see application references). Supplied as an agarose conjugate. Recognizes the ~94 kDa SV40 large T antigen.
Recognizes the ~94 kDa SV40 large T antigen in SV80 and SV-T2 cells. Cat. No. DP02A is linked to agarose for immunoprecipitation and purification.
This Anti-SV40 T Antigen (Ab-2) Mouse mAb (PAb416) Agarose Conjugate is validated for use in Immunoprecipitation, Affinity Purification for the detection of SV40 T Antigen (Ab-2).

Immunogen

purified SV40 large T-antigen
Epitope: Within amino acids 83-128

Application

Immunoprecipitation (15 µl suspension/sample)

Affinity Purification (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

50% slurry in PBS.

Analysis Note

Positive Control
SV80 or SV-T2 cells

Other Notes

DeCaprio, J.A., et al. 1988. Cell54, 275.
Whyte, P., et al. 1988. Nature334, 124.
Mitchell, P.J., et al. 1987. Cell50, 847.
Dixon, R.A.F. and Nathans, D., 1985. J. Virol.53, 1001.
Simanis, V. and Lane, D.P., 1985. Virol.144, 88.
Mann, R.S. and Carroll, R.B. 1984. Virology138, 379
Sarnow, P., et al. 1982. Cell28, 387.
Crawford, L.V., et al. 1981. Proc. Natl. Acad. Sci. USA78, 41.
Lane, D.P. and Crawford, L.V. 1979. Nature278, 261.
Carroll, R.B., et al. 1974. Proc. Natl. Acad. Sci. USA71, 3754.
Tooze, J.1973. Cold Spring Harbor, New York.
Tegtmeyer, P. 1972. J. Virol.10,
Suitable for affinity purification and immunoprecipitation. The amount of suspension required for purification is dependent on individual sample types.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NA02AOP03OP143
antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

species reactivity

SV40-infected cells

species reactivity

human, mouse, rat

species reactivity

rabbit, monkey, human, mouse, rat

species reactivity

mouse, human

clone

PAb416, monoclonal

clone

K121, monoclonal

clone

PAb421, monoclonal

clone

4B11, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

form

slurry (Liquid)

form

suspension (Liquid)

form

liquid

form

liquid


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
DP02A-0.5ML04055977226553

Questions

Reviews

No rating value

Active Filters