AC561
NovaSeptum® SURe Transfer Unit
with GO trigger and High Temperature AseptiQuik® S connector, sterile; γ-irradiated
About This Item
Recommended Products
material
polycarbonate connector
silicone tubing (
)
silicone tubing
Quality Level
sterility
sterile; γ-irradiated
product line
NovaSeptum® SURe
storage condition
room temperature
parameter
-80-125 °C temp. range (-12-257 °F)
0.5 bar max. pressure (7.25 psi)
fitting
inlet connection (Silicone)
outlet connection ( AseptiQuik® S HT)
application(s)
sterile sampling
General description
This product comes as a single unit with a NovaSeptum® GO trigger and connects via a High Temperature AseptiQuik® S connector. The design enable to autoclave your equipment and then connect any sampling design with an AseptiQuik® S connection. The closed design ensures your sample will be isolated from point of sample to analysis, reducing the risk of losing valuable product and maintaining the integrity of your fluid samples.
Application
Features and Benefits
- Product design ensures process integrity and a representative sample from single-use manufacturing processes
- Industry-standard aseptic connection and disconnection for easy integration into your single-use process
- Ideal for small volume sampling reducing waste of high value products
- Other container options available: bags, bottles, syringes, and conical tubes
- Stocked and ready-to-ship for fast delivery
Packaging
Preparation Note
Analysis Note
composed of materials which meet the criteria for
Biological Reactivity Testing.
These tests can be any one or a combination of the following test methods: USP <88> Class VI (in vivo), USP <87> (in vitro), ISO 10993-5 (in vitro).
Other Notes
Legal Information
also commonly purchased with this product
related product
required but not provided
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Learn more on mAb downstream processing, more specifically virus inactivation and the relevant associated products.
The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service