Skip to Content
Merck

Skip To

ABS984

Anti-Intersectin-1 Antibody

serum, from rabbit

Synonym(s):

ITSN-1, ITSN1, Intersectin-1, Intersectin1, ITSN 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITSN1(6453)

General description

Intersectin-1 is a scaffolding protein facilitating clathrin-mediated endocytosis, vesicle trafficking and mitogen-activated protein kinase (MAPK) signaling pathways. Functional association studies of the protein have suggested ITSN1 has several differing roles in various processes such as RAS activation and stimulation of Cdc42 in order to accelerate actin assembly. Alternative splicing produces 4 isoforms. The major isoforms are ITSN1-L and ITSN1-S. Expression for ITSN1 is ubiquitous in adult/fetal tissue, except isoform 1 which is mostly found in the brain. The most notable expression is found in muscle, spleen, skeletal muscle, testis, heart, ovary and all fetal tissue. Additionly, the neurons of young Down Syndrome and pre-Alzheimer’s disease brains are predisposed for enlarged early endosomes which may contribute to these pathological processes.
~150 and ~200 kDa observed. Previous publication indicated the 200 kDa to be the long isoform and 150 kDa to be the short isoform (Okur, M., et al. Mol. Cell. Biol. 2014, 34(2):271). Uncharacterized bands may be detected at high antibody concentrations or longer exposures.

Immunogen

Epitope: SH3 domain of Xenopus Intersectin-1 protein
GST-tagged recombinant protein corresponding to the SH3 domain of Xenopus Intersectin-1.

Application

Anti-Intersectin-1 is an antibody against Intersectin-1 for use in WB, ICC, IP.
Immunocytochemistry Analysis: A representative lot of this antibody detected Intersectin-1 protein in HEK293T cells (Martin, N., et al. Mol Pharmacol 70:1643–1653, 2006)
Immunoprecipitation Analysis: A representative lot of this antibody immunoprecipiated Intersectin-1 (short and long isoforms) from mouse brain lysate (Martin, N., et al. Mol Pharmacol 70:1643–1653, 2006)

Analysis Note

Evaluated by Western Blotting on rat brain cytosol tissue lysate

Western Blotting Analysis: A 1:1000 dilution of this antibody detected Intersectin-1 protein in 10 µg of rat brain cytosol.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA018007ABS983MABN894
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

antibody form

serum

antibody form

affinity isolated antibody

antibody form

serum

antibody form

purified antibody

Gene Information

human ... ITSN1(6453)

Gene Information

human ... ITSN1(6453)

Gene Information

human ... SLC11A2(4891)

Gene Information

human ... SYN1(6853)

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

10.22, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABS98404055977115796

Questions

Reviews

No rating value

Active Filters