Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 μg | Check Cart for Availability | € 277,00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse
species reactivity (predicted by homology)
human (immunogen homology)
technique(s)
immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... C1QTNF3(114899)
General description
Immunogen
Application
Immunofluorescence Analysis: 10 µg/mL from a representative lot detected CTRP3 in mouse heart cells.
Signaling
Developmental Signaling
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Mouse heart tissue lysate
Western Blotting/Peptide Inhibition Assay Analysis: 1 µg/mL of this antibody detected CTRP3 in 15 µg of mouse heart tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| species reactivity mouse | species reactivity human | species reactivity mouse, rat, human | species reactivity mouse, human |
| Gene Information human ... C1QTNF3(114899) | Gene Information human ... C1QTNF3(114899) | Gene Information human ... C1QTNF1(114897) | Gene Information human ... C1QTNF1(114897) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABS451 | 04053252879944 |



