Skip to Content
Merck

Skip To

ABS2269

Anti-TRPM4

Synonym(s):

Transient receptor potential cation channel subfamily M member 4;Calcium-activated non-selective cation channel 1;Long transient receptor potential channel 4;TrpC-4;LTrpC4;MLS2s;Melastatin-like 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 135.34 kDa, observed mol wt ~135 kDa

purified by

affinity chromatography

species reactivity

rat, mouse

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

C-terminal extracellular half

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

rat ... Trpm4(171143)

General description

Transient receptor potential cation channel subfamily M member 4 (UniProt: Q9ESQ5; also known as Calcium-activated non-selective cation channel 1, Long transient receptor potential channel 4, LTrpC-4, LTrpC4, MLS2s, Melastatin-like 2) is encoded by the Trpm4 (also known as Ltrpc4) gene (Gene ID: 171143) in rat. TRPM4 is a multi-pass membrane protein with four cytoplasmic domains; six transmembrane domains; and four extracellular domains. Two isoforms of TRPM4 have been described that are produced by alternative splicing. Isoform 1 is highly expressed in the testis tissue with a moderate expression in brain, spleen, and thymus. Isoform 2 is expressed in the brain and spleen. It serves as a calcium-activated non-selective cation channel that mediates membrane depolarization. Although it is a calcium activated channel, it is impermeable to calcium. It mediates the transport of monovalent cations that leads to membrane depolarization. It is reported to participates in T-cell activation by modulating Ca2+ oscillations after T lymphocyte activation, which is required for NFAT-dependent IL-2 production. TRPM4 is also involved in myogenic constriction of cerebral arteries. It plays a role in control of insulin secretion in pancreatic β-cells. It may also be involved in pacemaking activity or could cause irregular electrical activity under conditions of Calcium overload. It is known to affect Th1 and Th2 cell motility and cytokine production through differential regulation of calcium signaling and NFATC1 localization. This polyclonal antibody is reported toblock the channel vua binding to a region close to the channel porefrom extracellular space. Bu blocking TRPM4, it can downregulate TRPM4 surface expression and prevent hypoxia-induced cell swelling. (Ref.: Wei, S., et al. (2020). Front Cell Dev Bio. 8; 562584; Chen, B;et al. (2019). Pflugers Arch. 471(11-12); 1455-1466; Guinamard, R., et al. (2006). Hypertension. 48(4); 587-594; Launay, P., et al. (2002). Cell. 109(3); 397-407).

Immunogen

GST-conjugated linear peptide corresponding to 28 amino acids between transmembrane segment 5 and 6 from the C-terminal half of rat TRPM4.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells transfected with Mouse TRPM4.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected TRPM4 in lysate from HEK293 cells transfected with Mouse TRPM4.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected TRPM4 in Immunocytochemistry application (Low, S.W., et. al. (2021). Sci Rep. 11(1):10411).

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected TRPM4 in rat spinal cord tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

This rabbit polyclonal antibody detects Transient receptor potential cation channel subfamily M member 4 (TRPM4). It targets an epitope within 24 amino acids from the C-terminal, extracellular domain.

Physical form

Purified rabbit polyclonal antibody in PBS without azide.

Preparation Note

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABN418MABN748HPA041169
biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

species reactivity

rat, mouse

species reactivity

human, rat

species reactivity

mouse

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

clone

polyclonal

clone

polyclonal

clone

10H6, monoclonal

clone

polyclonal

Gene Information

rat ... Trpm4(171143)

Gene Information

human ... TRPM4(54795)

Gene Information

human ... TRPC3(7222)

Gene Information

human ... TRPM4(54795)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters