Skip to Content
Merck

Skip To

ABS22

Anti-PDE4D Antibody

from rabbit

Synonym(s):

phosphodiesterase 4D, cAMP-specific, cAMP-specific phosphodiesterase PDE4D6, cAMP-specific 3′,5′-cyclic phosphodiesterase 4D, phosphodiesterase 4D, cAMP-specific (dunce (Drosophila)-homolog phosphodiesterase E3), phosphodiesterase 4D, cAMP-specific (dunc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 397,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
6

€ 397,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

clone

polyclonal

species reactivity

rat, human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), mouse (based on 90% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDE4D(5144)

General description

Phosphodiesterase 4D (PDE4D) is a member of the PDE cyclic nucleotides. PDE4D is known to be regulated by protein kinase C (PKC)-Raf-MEK-ERK and also by cAMP in vascular smooth muscle cells. PDE4D is essential to have equilibrium in relaxing and contracting cues to airway smooth muscle. Down-regulation of PDE4D may be associated with analgesic and anti-inflammatory effects. It has also been linked to the risk of stroke, specifically common polygenic stroke.
~ 59, 68, 85, 90, and 105 kDa observed. UniProtKB/Swiss-Prot Q08499 (PDE4D_HUMAN) entry describes 12 isoforms produced by alternative splicing: Isoform 1 at 91.115 kDa, Isoform 2 at 76.369 kDa, Isoform 3 at 68.607 kDa, Isoform 4 at 66.376 kDa, Isoform 5 at 57.792 kDa, Isoform 6 at 84.428 kDa, Isoform 7 at 23.839 kDa, Isoform 8 at 59.113 kDa, Isoform 9 at 77.705 kDa, Isoform 10 at 76.816 kDa, Isoform 11 at 84.662 kDa, and Isoform 12 at 24.429 kDa

Immunogen

GST-tagged recombinant protein corresponding to the C-terminus of human PDE4D.

Application

Anti-PDE4D Antibody is an antibody against PDE4D for use in WB.

Biochem/physiol Actions

This antibody recognizes PDE4D at the C-terminus.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected PDE4D on 10 µg of rat brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABN117SAB4502128SAB2500765
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

goat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

ELISA: 1:10000, immunohistochemistry: 1:50-1:100, western blot: 1:500-1:1000

technique(s)

indirect ELISA: suitable, western blot: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABS2204053252520143

Questions

Reviews

No rating value

Active Filters