Skip to Content
Merck

Skip To

ABE480

Anti-Centrin-2 Antibody

serum, from rabbit

Synonym(s):

Centrin-2, Caltractin isoform 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 453,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, IP, WB
Citations:
2

€ 453,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CETN2(1069)

General description

Centrin-2 is also called Caltractin isoform 1. Centrin-2 is involved in microtubule organization, structure and function, and is required for centriole duplication and spindle formation. Centrin-2 also has a role in regulating cytokinesis, genome stability and global genome nucleotide excision repair (GG-NER) as component of the XPC complex.
~18 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Immunogen

Recombinant protein corresponding to human Centrin-2.

Application

Anti-Centrin-2 Antibody is a highly specific rabbit polyclonal antibody, that targets Centrin & has been tested in western blotting, ICC, IHC & IP.
Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Centrin-2 in MDCK cells (Koch, A., et al. (2010). Mol Biol Cell. 21(2):219-231.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Centrin-2 in wild type and galectin-3 null mouse kidney tissues (Koch, A., et al. (2010). Mol Biol Cell. 21(2):219-231.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Centrin-2 in MDCK cell lysate (Koch, A., et al. (2010). Mol Biol Cell. 21(2):219-231.).

Analysis Note

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Centrin-2 in 10 µg of Jurkat cell lysate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1405608C7736SAB5701604
antibody form

serum

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

-

Quality Level

200

Quality Level

100

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

UniProt accession no.

P41208

UniProt accession no.

P41208

UniProt accession no.

Q12798

UniProt accession no.

P41208

species reactivity

human, mouse, rat

species reactivity

human

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ting Zhang et al.
Nature communications, 13(1), 6907-6907 (2022-11-15)
Transcription replication collisions (TRCs) constitute a major intrinsic source of genome instability but conclusive evidence for a causal role of TRCs in tumor initiation is missing. We discover that lack of the H4K20-dimethyltransferase KMT5B (also known as SUV4-20H1) in muscle
Renee A Lopez et al.
Bio-protocol, 7(21), e2599-e2599 (2017-11-05)
Male gametes (spermatozoids) are the only motile cells produced during the life cycle of land plants. While absent from flowering and most cone-bearing plants, motile cells are found in less derived taxa, including bryophytes (mosses, liverworts and hornworts), pteridophytes (lycophytes



Global Trade Item Number

SKUGTIN
ABE48004053252913938

Questions

Reviews

No rating value

Active Filters