Skip to Content
Merck

Skip To

ABE367

Anti-RNF168 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

E3 ubiquitin-protein ligase RNF168, hRNF168, RING finger protein 168, RNF168

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 453,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
24

€ 453,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RNF168(165918)

General description

E3 ubiquitin-protein ligase RNF168 (UniProt Q8IYW5; also known as hRNF168, RING finger protein 168) is encoded by the RNF168 gene (Gene ID 165918) in human. Ubiquitylation/Ubiquitination is an essential early signal in the DNA damage repair process. Multiple E3 ligases, including RNF2 (RING2), RNF8 and RNF168, mediate the addition of K63-linked polyubiquitin chains to histone 2A (H2A) and H2AX, which in turn recruits DNA damage repair proteins, e.g., RAP80, BRCA1. In addition, SUMOylation and NEDDylation are also involved in the DNA damage response. RNF168 is a dual specificity E3 ligase that mediates both the ubiquitination and NEDDylation of H2A and H2AX. The two types of modifications compete against each other and NEDD8 modification of H2A and H2AX blocks the recruitment of BRCA1 at DNA damage repair sites. RNF168 itself is also subjected to NEDD8 modification and NEDDylation of RNF168 is necessary for its ubiquitin ligase activity. Inhibition of RNF168 NEDDylation impairs its interaction with E2 enzyme Ubc13 (UBE2N). Likewise, downregulating RNF168 NEDDylation by the deNEDDylating enzyme NEDP1, or due to mutations of the NEDD8-conjugating enzyme UBC12, decreases H2A and H2AX ubiquitylation. RNF168 Mutations are linked to RIDDLE (radiosensitivity, immunodeficiency dysmorphic features and learning difficulties) syndrome.
~75 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: C-terminus
GST-tagged recombinant human RNF168.

Application

Immunocytochemistry Analysis: 1.0 µg/mL from a representative lot detected RNF168 in HeLa, A431, HUVEC, and NIH/3T3 cells.
Immunohistochemistry Analysis: A representative lot detected RNF168 in frozen human lung adenocarcinoma tissue sections (Xie, X., et al. (2018) Nat. Cell Biol. 20(3); 320-331).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Use Anti-RNF168 Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC, IF to detect RNF168 also known as E3 ubiquitin-protein ligase RNF168.

Biochem/physiol Actions

This antibody detects RNF168 in a lymphoblastoid cell line (LCL) derived from a healthy donor, but not LCL derived from a RIDDLE (radiosensitivity, immunodeficiency dysmorphic features and learning difficulties) syndrome patient.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blotting in SA13 human B cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RNF168 in 8 µg of SA13 human B cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1410293SAB1408409WH0165918M1
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

Gene Information

human ... RNF168(165918)

Gene Information

human ... RNF168(165918)

Gene Information

human ... RNF168(165918)

Gene Information

human ... RNF168(165918)

species reactivity

human, mouse

species reactivity

human

species reactivity

human

species reactivity

human, mouse, rat


Still not finding the right product?

Explore all of our products under Anti-RNF168 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins


Wenwen Wu et al.
Cancer science, 107(10), 1406-1415 (2016-10-30)
The breast and ovarian cancer predisposition protein BRCA1 forms three mutually exclusive complexes with Fanconi anemia group J protein (FANCJ, also called BACH1 or BRIP1), CtIP, and Abraxas/RAP80 through its BRCA1 C terminus (BRCT) domains, while its RING domain binds
Xiaoduo Xie et al.
Nature cell biology, 20(3), 320-331 (2018-02-07)
Growth signals, such as extracellular nutrients and growth factors, have substantial effects on genome integrity; however, the direct underlying link remains unclear. Here, we show that the mechanistic target of rapamycin (mTOR)-ribosomal S6 kinase (S6K) pathway, a central regulator of
I K Mandemaker et al.
Scientific reports, 7(1), 15353-15353 (2017-11-12)
The DNA damage response (DDR), comprising distinct repair and signalling pathways, safeguards genomic integrity. Protein ubiquitylation is an important regulatory mechanism of the DDR. To study its role in the UV-induced DDR, we characterized changes in protein ubiquitylation following DNA



Global Trade Item Number

SKUGTIN
ABE36704053252537219

Questions

Reviews

No rating value

Active Filters