Skip to Content
Merck

Skip To

ABE302

Anti-ATDC Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ataxia telangiectasia group D-associated protein, tripartite motif containing protein 29

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRIM29(23650)

General description

Ataxia telangiectasia group D-complementing (ATDC) protein, also known as tripartite motif containing protein 29 (TRIM29), belongs to the TRIM protein family. Studies have shown that ATDC may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. The ATDC gene is located at 11q23 where loss of heterozygosity has been observed in many kinds of cancers; underexpression of ATDC has been reported in breast and prostate cancer. ATDC may also complement the radiosensitivity defect of ataxia telangiectasia phenotype.
~66 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to a region within amino acides 565 to 595 of human ATDC .

Application

Anti-ATDC Antibody is a Rabbit Polyclonal Antibody for detection of ATDC also known as Ataxia telangiectasia group D-associated protein & has been validated in WB & IP.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in IP. (Yuan, Z., et al. (2010). J Biol Chem. 285(50):39329-39338.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
RAW 264.7 cell lysate
Evaluated by Western Blot in RAW 264.7 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected ATDC on 10 µg of RAW 264.7 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA020053SAB2501512ABE208
biological source

rabbit

biological source

rabbit

biological source

goat

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

UniProt accession no.

Q14134

UniProt accession no.

Q14134

UniProt accession no.

Q14134

UniProt accession no.

P35222

species reactivity

human, mouse, rat

species reactivity

human, rat

species reactivity

human, mouse, rat

species reactivity

mouse, human, rat, horse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters