Skip to Content
Merck

Skip To

ABE241

Anti-MBNL1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

muscleblind-like (Drosophila), Triplet-expansion RNA-binding protein, muscleblind (Drosophila)-like, muscleblind-like protein 1, KIAA0428

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 428,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
2

€ 428,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1150

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MBNL1(4154)

General description

Muscleblind 1 (MBNL1) is a member of the muscleblind family. MBNL1in involved in pre-mRNA alternative splicing and acts as either splicing activator or repressor on specific pre-mRNA targets. This protein contains four C3H1-type zinc fingers and binds to expanded CUG repeat RNA. High expression of MBNL1 has been observed in cardiac and skeletal muscle as well as in differentiating myoblasts. MBNL1 is known to be involved in the pathogenesis of dystrophia myotonica type 1 (DM1). For those afflicted with this muscular disorder, MBNL1 is sequestered by DMPK RNAs that contain CUG triplet repeat expansions.
~ 33-42 kDa observed. There are three known splice variants ranging from ~33 kDa to ~42 kDa An uncharacterized band at ~ 80 kDa has been observed in some lysates.

Immunogen

Histidine-tagged recombinant protein corresponding to human MBNL1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-MBNL1 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect MBNL1 also known as muscleblind-like (Drosophila), Triplet-expansion RNA-binding protein.

Physical form

Affinity purified
Purified rabbit polyclonal in tris-citrate containing 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate
Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected MBNL1 on 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV41228SAB1404054MABE959
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse, human, rat

species reactivity

human

species reactivity

human

species reactivity

mouse

UniProt accession no.

Q9NR56

UniProt accession no.

Q9NR56

UniProt accession no.

Q9NR56

UniProt accession no.

Q5VZF2


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABE24104053252642531

Questions

Reviews

No rating value

Active Filters