About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse
species reactivity (predicted by homology)
frog (based on 100% sequence homology), bovine (based on 100% sequence homology), chicken (based on 100% sequence homology), human (immunogen homology)
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ZEB1(6935)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Neuroscience
Transcription Factors
Developmental Signaling
Physical form
Preparation Note
Analysis Note
Mouse embryo e11 tissue lysate
Western Blot Analysis: 1.0 µg/mL of this antibody detected ZEB1 in 10 µg of mouse embryo e11 tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source mouse | biological source rabbit |
| technique(s) western blot: suitable | technique(s) immunofluorescence: suitable, western blot: suitable | technique(s) immunofluorescence: suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL | technique(s) immunohistochemistry: suitable, western blot: suitable |
| Gene Information human ... ZEB1(6935) | Gene Information human ... ZEB1(6935) | Gene Information human ... ZEB1(6935) | Gene Information human ... ZEB2(9839) |
Still not finding the right product?
Explore all of our products under Anti-ZEB1 Antibody
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABD53 | 04053252832475 |



