Skip to Content
Merck

Skip To

ABD121

Anti-PRDM14 Antibody

from rabbit

Synonym(s):

PRDM-14, PR domain zinc finger protein 14

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, WB
Citations:
1


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ChIP: suitable (ChIP-seq), western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRDM14(63978)

General description

PRDM14 is an approximately 65 kDa transcriptional inhibitor that contains one histone methylating PR/SET domain (aa 266 - 367) and six zinc finger repeats (aa 400 - 568). PRDM14 is preferentially expressed in germ cells and undifferentiated embryonic stem cells in which it plays a central role in lineage specification, maintenance of pluripotency, and repression of somatic gene transcription. PRDM14 is upregulated in breast cancer and lymphoblastic leukemia. Recently, PRDM14 ensures naive pluripotency through dual regulation of signaling and epigenetic pathways in embryonic stem cells

~64 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: N-terminus
Human recombinant PRDM14.

Application

Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory on H1 human ES cells (CHAN et al., 2013. STEM CELLS, 31:682–692)

ChIPseq Analysis: A representative lot was used by an independent laboratory on H1 human ES cells (Chia et al., 2010. Nature. 316, VOL 468)
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Anti-PRDM14 Antibody is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of PRDM14 .

Biochem/physiol Actions

Human

Physical form

Format: Unpurified
Raw rabbit polyclonal serum
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in Human ES cell cell lysate. Western Blot Analysis: 1 µg/mL of this antibody detected PRDM14 in undifferentiated human ES cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB4350ABD130ABE543
antibody form

unpurified

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Gene Information

human ... PRDM14(63978)

Gene Information

human ... PRDM14(63978)

Gene Information

human ... PRDM16(63976)

Gene Information

human ... PRDM16(63976)

species reactivity

human

species reactivity

rat, mouse, human

species reactivity

rat, human

species reactivity

human


Still not finding the right product?

Explore all of our products under Anti-PRDM14 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Cancer is a complex disease manifestation. At its core, it remains a disease of abnormal cellular proliferation and inappropriate gene expression. In the early days, carcinogenesis was viewed simply as resulting from a collection of genetic mutations that altered the gene expression of key oncogenic genes or tumor suppressor genes leading to uncontrolled growth and disease (Virani, S et al 2012). Today, however, research is showing that carcinogenesis results from the successive accumulation of heritable genetic and epigenetic changes. Moreover, the success in how we predict, treat and overcome cancer will likely involve not only understanding the consequences of direct genetic changes that can cause cancer, but also how the epigenetic and environmental changes cause cancer (Johnson C et al 2015; Waldmann T et al 2013). Epigenetics is the study of heritable gene expression as it relates to changes in DNA structure that are not tied to changes in DNA sequence but, instead, are tied to how the nucleic acid material is read or processed via the myriad of protein-protein, protein-nucleic acid, and nucleic acid-nucleic acid interactions that ultimately manifest themselves into a specific expression phenotype (Ngai SC et al 2012, Johnson C et al 2015). This review will discuss some of the principal aspects of epigenetic research and how they relate to our current understanding of carcinogenesis. Because epigenetics affects phenotype and changes in epigenetics are thought to be key to environmental adaptability and thus may in fact be reversed or manipulated, understanding the integration of experimental and epidemiologic science surrounding cancer and its many manifestations should lead to more effective cancer prognostics as well as treatments (Virani S et al 2012).


Anastasiya Sybirna et al.
Nature communications, 11(1), 1282-1282 (2020-03-11)
PRDM14 is a crucial regulator of mouse primordial germ cells (mPGCs), epigenetic reprogramming and pluripotency, but its role in the evolutionarily divergent regulatory network of human PGCs (hPGCs) remains unclear. Besides, a previous knockdown study indicated that PRDM14 might be



Global Trade Item Number

SKUGTIN
ABD12104053252993039

Questions

Reviews

No rating value

Active Filters