Skip to Content
Merck

Skip To

ABC428

Anti-VEGF-D Antibody

1 mg/mL, from rabbit

Synonym(s):

Vascular endothelial growth factor D, VEGF-D, c-Fos-induced growth factor, FIGF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 339,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

€ 339,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

concentration

1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FIGF(2277)

General description

VEGF-D, also known as Vascular Endothelial Growth Factor D, or c-Fos-induced growth factor, FIGF, and encoded by the gene FIGF/VEGFD, is one of a family of vascular growth factors that is active in angiogenesis, lymphangiogenesis and endothelial cell growth. VEGF-D functions during the formation of venous and lymphatic vascular systems and is critical for their maintenance during adulthood. VEGF-D, originally discovered as a c-Fos induced growth factor (FIGF), is developmentally regulated by c-Fos and expressed in a variety of tissues and organs including limb buds, acoustic ganglion, teeth, heart, anterior pituitary as well as lung and kidney mesenchyme, liver, derma, and periosteum of the vertebral column. Because of its role in lymphangiogenesis VEGFD is also a key therapeutic target to modulate metastasis, and recent research demonstrates that VEGFD-MEK-ERK signaling modulates SOX18-mediated transcription in normal cells and thus various pathologies maybe because of a dysregulation of SOXF-mediated transcriptional networks.
~57 kDa observed. The calculated molecular weight is 40 kDa, however VEGF-D has been shown as a ~57 to ~58 kDa band in western blots (Liu Y.H., et al., (2008) J Immunol. 81(9):6584-94).

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human VEGF-D near the N-terminus.

Application

Anti-VEGF-D Antibody detects level of VEGF-D & has been published & validated for use in VEGF-D.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in SW620 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected VEGF-D in 10 µg of SW620 cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1306333SAB1405812ABC429
antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

Gene Information

human ... FIGF(2277)

Gene Information

human ... FIGF(2277)

Gene Information

human ... FIGF(2277)

Gene Information

human ... VEGFB(7423)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABC42804053252977497

Questions

Reviews

No rating value

Active Filters