Skip to Content
Merck

Skip To

ABC114

Anti-Thrombospondin-2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Thrombospondin-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 358,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

€ 358,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... THBS2(7058)

General description

Thrombospondins (TSPs) are a family of homotrimeric multi-domain glycoproteins involved in modulation of cell adhesion, proliferation, and migration and have been implicated clinically in the development of atherosclerosis, angiogenesis, and oncogenesis. TSP2, a member of the thrombospondin family is a secreted protein, but is present only transiently in extracellular matrix and is rapidly internalized for degradation by fibroblasts after binding to the cell surface. It also interacts with matrix metalloproteinases (MMPs) and matrix serine proteases, such as plasminogen activator. TSP2 is reportedly controls the apoptosis-proliferation balance in endothelial cells. Although TSP2 is a potent inhibitor of angiogenesis, its N-terminal region exhibits pro-angiogenic activities mediated by beta 1 integrins. Loss of TSP2 expression is associated with local invasive behavior, tumor neovascularization, and metastasis. Decreased TSP2 expression has been correlated with malignant progression and angiogenesis in some cancers. TSP2 levels are also highly expressed in developing and healing tissues.
~180 kDa observed. The calculated molecular weight is 130 kDa, however Thrombospondin-2 has been shown to have bands at ~135 kDa and ~180 kDa in western blots (Streit, M., et al. (1999). PNAS. 96(26):14888-14893).

Immunogen

Epitope: Laminin G-like region
KLH-conjugated linear peptide corresponding to the Laminin G-like region of human Thrombospondin-2.

Application

Anti-Thrombospondin-2 Antibody is an antibody against Thrombospondin-2 for use in Western Blotting.

Biochem/physiol Actions

Other homologies: Mouse, Rat, and Bovine (93% sequence homology).
This antibody recognizes the Laminin G-like region of Thrombospondin-2.

Analysis Note

Control
Concentrated conditioned media from MG-63 cells
Evaluated by Western Blot in concentrated conditioned media from MG-63 cells.

Western Blot Analysis: 0.25 µg/mL of this antibody detected Thrombospondin-2 in 10 µg of concentrated conditioned media from MG-63 cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABT879SAB5500039MAB4444
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

mouse

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

technique(s)

western blot: suitable

technique(s)

ELISA: suitable, immunohistochemistry: suitable (paraffin), neutralization: suitable, western blot: suitable

technique(s)

immunohistochemistry: 1:100

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable

species reactivity

human

species reactivity

rat, human

species reactivity

human (tested)

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters