Skip to Content
Merck

Skip To

AB9983

Anti-Tyrosine Hydroxylase Antibody

rabbit polyclonal

Synonym(s):

Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 401,00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
polyclonal
Species reactivity:
rat, mouse
Application:
IHC, WB
Citations:
10

€ 401,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1033

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Tyrosine Hydroxylase Antibody, from rabbit

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Th(21823)
rat ... Th(25085)

General description

Tyrosine Hydroxylase plays an important role in the physiology of adrenergic neurons. It is the first and rate-limiting enzyme involved in the biosynthesis of the catecholamines Dopamine and Norepinephrine from tyrosine. TH is, therefore, a useful marker for dopaminergic and noradrenergic neurons. The enzymatic activity of TH requires ferrous ions as cofactors and is believed to be regulated by phosphorylation. At least four isoforms of human TH have been identified which result from alternative splicing.
~ 58 kDa

Immunogen

Epitope: C-terminus
Recombinant protein

Application

Anti-Tyrosine Hydroxylase Antibody detects level of Tyrosine Hydroxylase & has been published & validated for use in IH(P) & WB.
Immunohistochemistry: 1:100 working dilutions were used on rat and mouse brain (paraffin embedded tissue).
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Biochem/physiol Actions

Cat. # AB9983 recognizes the C-terminus of Tyrosine Hydroxylase
Reactivity with other species has not been determined.

Physical form

Antigen Affinity Purified
Format: Purified
Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3

Preparation Note

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Rat Brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB1542AB152AB5986
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

serum

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

sheep

biological source

rabbit

biological source

rabbit

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Gene Information

mouse ... Th(21823)
rat ... Th(25085)

Gene Information

human ... TH(7054)

Gene Information

human ... TH(7054)
rat ... Th(25085)

Gene Information

rat ... Th(25085)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunohistochemistry: suitable


Still not finding the right product?

Explore all of our products under Anti-Tyrosine Hydroxylase Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Stefano Garofalo et al.
Nature communications, 14(1), 3103-3103 (2023-05-30)
The mechanisms of communication between the brain and the immune cells are still largely unclear. Here, we characterize the populations of resident natural killer (NK) cells and innate lymphoid cells (ILC) 1 in the meningeal dura layer of adult mice.
Qiong L Zhou et al.
Molecular and cellular biology, 38(8) (2018-01-31)
CDP138 is a calcium- and lipid-binding protein that is involved in membrane trafficking. Here, we report that mice without CDP138 develop obesity under normal chow diet (NCD) or high-fat diet (HFD) conditions. CDP138-/- mice have lower energy expenditure, oxygen consumption
Krishnan Sriram et al.
Toxicology and applied pharmacology, 449, 116137-116137 (2022-06-25)
Workers in the oil and gas industry are at risk for exposure to a number of physical and chemical hazards at the workplace. Chemical hazard risks include inhalation of crude oil or its volatile components. While several studies have investigated



Global Trade Item Number

SKUGTIN
AB998304053252409738

Questions

Reviews

No rating value

Active Filters