Skip to Content
Merck

Skip To

AB9486

Anti-Somatostatin Receptor Type 2 Antibody

Chemicon®, from rabbit

Synonym(s):

SSTR2

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC
Citations:
-


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SSTR2(6752)

Immunogen

Synthetic peptide from the 2nd extracellular domain of human Somatostatin Receptor Type 2.

Application

Detect Somatostatin Receptor Type 2 using this Anti-Somatostatin Receptor Type 2 Antibody validated for use in IH(P).
Immunohistochemistry on paraffin embedded tissue sections.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors

Biochem/physiol Actions

Recognizes Somatostatin Receptor Type 2 [SSTR2].

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.

Preparation Note

Maintain at -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S1445AB9287HPA007264
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

Gene Information

human ... SSTR2(6752)

Gene Information

human ... SSTR1(6751)
mouse ... Sstr1(20605)
rat ... Sstr1(25033)

Gene Information

human ... SSTR3(6753)

Gene Information

human ... SSTR2(6752)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters