Skip to Content
Merck

Skip To

AB2256

Anti-GABA B Receptor R1 Antibody

serum, from guinea pig

Synonym(s):

GABA-B receptor, GABA-B receptor 1, GABAB, subunit 1c, gamma-aminobutyric acid (GABA) B receptor 1, gamma-aminobutyric acid B receptor 1, seven transmembrane helix receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 μL
Check Cart for Availability
€ 504,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
9

€ 504,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

guinea pig

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Gabbr1(54393)

General description

Gamma-aminobutyric acid (GABA) is the main inhibitory neurotransmitter in the mammalian central nervous system. GABA exerts its effects through ionotropic [GABA(A/C)] receptors, to produce fast synaptic inhibition, and metabotropic [GABA(B)] receptors, to produce slow, prolonged inhibitory signals. The GABA(B) receptor consists of a heterodimer of two related 7-transmembrane receptors, GABA(B) receptor 1 and GABA(B) receptor 2. The GABA(B) receptor 1 gene is mapped to chromosome 6p21.3 within the HLA class I region close to the HLA-F gene. Susceptibility loci for multiple sclerosis, epilepsy, and schizophrenia have also been mapped in this region. Alternative splicing of this gene generates 4 transcript variants.
~100-105 kDa

Immunogen

Epitope: C-Terminus
interaction w/ ATF4
Recombinant peptide (aa 972-991: PSEPPDRLSCDGSRVHLLYK)

Application

Detect GABA B Receptor R1 using this Anti-GABA B Receptor R1 Antibody validated for use in WB, IH.
Immunohistochemistry: Working dilution of 1:3,000 in free floating mouse brain tissue.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Biochem/physiol Actions

Reactivity with other species has not been determined.
Cat. # AB2256 recognizes GABA B Receptor R1 (GABA B R1)

Physical form

GST-Depleted Sera with 0.05% NaN3.

Preparation Note

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Rat brain lysates
Western Blotting: 1:1,000 dilution from a previous lot detected GABA B R1 on 10µg of rat brain lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4501062SAB4501063MABN263
antibody form

serum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

guinea pig

biological source

rabbit

biological source

rabbit

biological source

mouse

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

10F10-C1-B8, monoclonal

Gene Information

mouse ... Gabbr1(54393)

Gene Information

human ... GABBR1(2550)

Gene Information

human ... GABBR1(2550)

Gene Information

rat ... Gabrg2(29709)


Still not finding the right product?

Explore all of our products under Anti-GABA B Receptor R1 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
AB225604053252513848

Questions

Reviews

No rating value

Active Filters