Skip to Content
Merck

Skip To

AB1975

Anti-Enkephalin, Met Antibody

serum, Chemicon®

Synonym(s):

Anti-PE, Anti-PENK-A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μL
Check Cart for Availability
€ 424,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC
Citations:
3

€ 424,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

suitability

not suitable for Western blot

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

rat ... Penk(29237)

General description

0.6 kDa
Met Enkephalin is an endogenous opioid peptide neurotransmitter found naturally in the brains of many animals, including humans. Methionine enkephalin (Met Enkephalin) is a pentapeptide that appears to enhance immune function at low doses and suppress at high doses. This molecule acts in the central nervous, neuroendocrine, and immune systems. During development of the brain Met Enkephalin is thought to act as a growth factor. Met Enkephalin may be capable of enhancing immune function in cancer or AIDS patients.

Immunogen

Met-enkephalin, Hemocyanin conjugate.

Application

Immunohistochemistry (with immunoperoxidase detection *): 48 hour incubation, 4°C.

Spinal cord nerve fibers (rat): 1:2,000-1:4,000.

Immunofluorescence*: overnight incubation, +2-8°C

Duodenum endocrine cells & nerve fibers (pig): 1:2,000

Ileum endocrine cells & nerve fibers μguinea pig): 1:4,000.

*fixative-any cross-linking reagent

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Hormones & Receptors
This Anti-Enkephalin, Met Antibody is validated for use in IH for the detection of Enkephalin.

Biochem/physiol Actions

Met-enkephalin

Physical form

Serum in 0.05% Sodium Azide

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Tested on rat brain/coronal section

Other Notes

Replaces: AB5026

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M5795A2066SAB2500825
antibody form

serum

antibody form

whole antiserum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

goat

species reactivity

rat

species reactivity

mouse, human, rat

species reactivity

wide range, vertebrates, human, slime mold, amoeba, chicken

species reactivity

human

UniProt accession no.

P01210

UniProt accession no.

P36507

UniProt accession no.

P68133

UniProt accession no.

P01210


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters