Skip to Content
Merck

Skip To

AB15554

Anti-AT2 Receptor Antibody

Chemicon®, from rabbit

Synonym(s):

Angiotensin II receptory type 2, AGTR2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μL
Check Cart for Availability
€ 467,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
6

€ 467,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

should not react with

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AGTR2(186)

Immunogen

Purified peptide from the N-terminus of rat AT2 Receptor (Accession number P35351).

Application

This Anti-AT2 Receptor Antibody is validated for use in WB for the detection of AT2 Receptor.
Western blot: 1:200-1:500 using ECL on rat brain and mouse MS1 cell lysate.
Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Recognizes AT2 Receptor (Angiotensin II receptory type 2, AGTR2).

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB15552SAB2500038SAB5700087
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

goat

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Gene Information

human ... AGTR2(186)

Gene Information

human ... AGTR1(185)

Gene Information

human ... AGTR1(185)

Gene Information

human ... AGTR1(185)

technique(s)

western blot: suitable

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

indirect ELISA: suitable, western blot: suitable

technique(s)

western blot: 1:500-1:2000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters