The maximum temperature that this resin can withstand has not been determined. Upon reviewing available literature, one citation reports usage of Wang resin at 40–50°C. Please see the link below to review this publication:
https://www.sciencedirect.com/topics/chemistry/wang-resin
Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 5 g | Check Cart for Availability | € 176,00 | |
| 25 g | Check Cart for Availability | € 702,00 | |
| 100 g | Check Cart for Availability | € 2.630,00 |
About This Item
Quality Segment
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
storage temp.
2-8°C
General description
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Literature references
[1] C. D. Floyd, et al. (1996) Tetrahedron Lett., 37, 8045.
[2] D. E. Robinson & M. W. Holladay (2000) Org. Lett., 2, 2777.
[3] P. Blaney, et al. (2000) Tetrahedron Lett., 41, 6635.
[4] A. Volonterio, et al. (2002) Eur. J. Org. Chem., 428.
[5] P. Blaney, et al. (2000) Tetrahedron Lett., 41, 6639.
Analysis Note
Appearance of substance (visual): beads
Loading (determined by elemental analysis of nitrogen): 1.5 - 2.5 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene - 1% DVB), 100 - 200 mesh.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis |
| product line Novabiochem® | product line Novabiochem® | product line - | product line Novabiochem® |
| Quality Level 200 | Quality Level 400 | Quality Level 100 | Quality Level 200 |
| application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis |
| storage temp. 2-8°C | storage temp. 2-30°C | storage temp. - | storage temp. 2-30°C |
| form beads | form beads | form solid | form beads |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8551170005 | 04027269114619 |
| 8551170025 | 04027269114626 |
| 8551170100 | 04027269114633 |
-
What is the maximum temperature that this resin can withstand?
1 answer-
Helpful?
-
-
Does this Wang resin have a Fmoc group on it? Do I need to remove it when I first use it? Thanks so much!
1 answer-
This resin doesn't contain a Fmoc group but it can be added. Please refer to the following article, https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/peptide-synthesis/peptide-resin-loading.
Helpful?
-



