Skip to Content
Merck

Skip To

8.55116

Methyl Indole AM resin

Novabiochem®

Synonym(s):

AM Methylindole Resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 106,00
€ 79,50
5 g
Check Cart for Availability
€ 427,00
25 g
Check Cart for Availability
€ 1.610,00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352005

€ 79,50

List Price€ 106,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

[3-((Methyl-Fmoc-amino)-methyl)indol-1-yl] acetyl AM resin is an extremely useful support for the production of N-methyl substituted carboxamides and sulfonamides, as well as peptide N-methylamides [1]. After first removing the Fmoc group, loading of the resin with Fmoc-amino acids can be effected using HATU/DIPEA. Cleavage with TFA directly provides the product N-methylamide.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins


Literature references

[1] K. G. Estep, et al. (1998) J. Org. Chem., 63, 5300.

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.60 - 1.00 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 100 - 200 mesh.

Other Notes

Replaces: 01-64-0451

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.551028.550288.55134
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

2-8°C

form

beads

form

beads

form

beads

form

beads


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Will TFA treatment of peptides synthesized on the Methyl Indole AM resin result in peptides with methyl amide c-terminals?

    1 answer
    1. This resin is an extremely useful support for the production of N-methyl substituted carboxamides and sulfonamides, as well as peptide N-methylamides.

      After first removing the Fmoc group with piperidine, loading of the resin with Fmoc-amino acids can be effected using HATU/DIPEA. Cleavage with TFA directly provides the product N-methylamide. Please refer to method 4 for more information on a loading protocol for attachment of carboxylic acids to amino resins and method 2 for more information on general TFA cleavage.

      Method 4 : https://www.sigmaaldrich.com/technical-documents/technical-article/chemistry-and-synthesis/peptide-synthesis/peptide-resin-loading#Method4

      Method 2 : https://www.sigmaaldrich.com/technical-documents/technical-article/chemistry-and-synthesis/peptide-synthesis/fmoc-cleavage-deprotection#Method2

      Helpful?

Reviews

No rating value

Active Filters