Skip to Content
Merck
All Photos(1)

Key Documents

8.55073

Sigma-Aldrich

NovaSyn TG amino resin HL

Novabiochem®

Synonym(s):

NovaSyn TG amino resin HL

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€75.40
5 G
€308.00

€75.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€75.40
5 G
€308.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352203
NACRES:
NA.22

€75.40


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

General description

High load version of NovaSyn TG amino resin.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Linkage

Replaces: 01-64-0144

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.30 - 0.60 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
This resin is a composite of low-crossed linked polystyrene and 3000 - 4000 M.W. polyethyleneglycol, which has been terminally amino functionalised. 110 µm beads.

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers polymer supports for solid phase peptide synthesis, suitable for various peptide lengths and sequences.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service