Skip to Content
Merck
All Photos(1)

Key Documents

86359

Sigma-Aldrich

TentaGel S-NH2

extent of labeling: 0.20-0.35 mmol/g N loading

Synonym(s):

O-(2-Aminoethyl)polyethylene glycol polymer-bound

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€214.00

€214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€214.00

About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€214.00


Check Cart for Availability

Request a Bulk Order

product line

TentaGel

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.20-0.35 mmol/g N loading

particle size

110-150 μm

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

TentaGel S-NH2 can be used:
  • As a support in the synthesis of L-Leu based peptides and oligomers, which are used as catalysts in Julia-Colonna epoxidation of chalcones.[1]
  • In the synthesis of 1,3-dialkyl quinazoline-2,4-diones.[2]
  • To synthesize N-substituted amidinoaryloxypyridine analogs as potent factor Xa inhibitors.[3]

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly enantioselective enone epoxidation catalyzed by short solid phase-bound peptides: dominant role of peptide helicity
Berkessel A, et al.
Organic Letters, 3(24), 3839-3842 (2001)
Solid-phase synthesis of 1, 3-dialkyl quinazoline-2, 4-diones
Buckman BO and Mohan R
Tetrahedron Letters, 37(26), 4439-4442 (1996)
Solid-phase synthesis of N-substituted amidinophenoxy pyridines as factor Xa inhibitors
Mohan R, et al.
Bioorganic & Medicinal Chemistry Letters, 8(14), 1877-1882 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service