Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 5 g | Check Cart for Availability | € 381,00 |
About This Item
Quality Level
product line
Novabiochem®
assay
≥97% (TLC), ≥97.0% (HPLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
hydroxyl
storage temp.
2-8°C
SMILES string
N5([C@@H]([C@H](OC5(C)C)C)C(=O)O)C(=O)[C@@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)Cc1ccc(cc1)OC(C)(C)C
InChI
1S/C35H40N2O7/c1-21-30(32(39)40)37(35(5,6)43-21)31(38)29(19-22-15-17-23(18-16-22)44-34(2,3)4)36-33(41)42-20-28-26-13-9-7-11-24(26)25-12-8-10-14-27(25)28/h7-18,21,28-30H,19-20H2,1-6H3,(H,36,41)(H,39,40)/t21-,29+,30+/m1/s1
InChI key
JYSMURJYNLWZNY-RIGQTMPJSA-N
General description
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS
Analysis Note
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -32.0 - -26.0 °
Purity (TLC(157A)): ≥ 97 %
Purity (TLC(CMA2)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Single impurities (HPLC, area%): ≤ 1.00 % (a/a)
Ethyl acetate (HS-GC): ≤ 2.0 %
Acetate (IC): ≤ 0.1 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form powder | form powder | form powder | form powder |
| assay ≥97% (TLC), ≥97.0% (HPLC) | assay ≥97% (TLC), ≥97.0% (HPLC) | assay ≥97% (TLC), ≥97.0% (HPLC) | assay ≥97% (TLC), ≥97.0% (HPLC) |
| functional group hydroxyl | functional group hydroxyl | functional group hydroxyl | functional group hydroxyl |
| product line Novabiochem® | product line Novabiochem® | product line Novabiochem® | product line Novabiochem® |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
The ease of assembly of a given peptide sequence is hard to predict, which makes peptide synthesis challenging. Review methods and reagents for avoiding aggregation in solid-phase peptide synthesis.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8521820025 | 04027269190316 |
| 8521820005 | 04027269092719 |
| 8521820001 | 04027269092702 |
| 8521820100 | 04027269190293 |



