Skip to Content
Merck

Skip To

8.52154

Fmoc-D-Asp(OtBu)-OH

≥99.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-D-Asp(OtBu)-OH, N-α-Fmoc-D-aspartic acid β-t.-butyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H25NO6
CAS Number:
Molecular Weight:
411.45
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fmoc-D-Asp(OtBu)-OH, Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥96.0% (acidimetric), ≥98% (TLC), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

2-30°C

SMILES string

N([C@H](CC(=O)OC(C)(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C23H25NO6/c1-23(2,3)30-20(25)12-19(21(26)27)24-22(28)29-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,24,28)(H,26,27)/t19-/m1/s1

InChI key

FODJWPHPWBKDON-LJQANCHMSA-N

General description

Standard building block of introduction of D-aspartic acid amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 96.0 %
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-13-1050

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.520058.521448.52004
form

powder

form

powder

form

powder

form

powder

functional group

carboxylic acid

functional group

carboxylic acid

functional group

carboxylic acid

functional group

carboxylic acid

assay

≥96.0% (acidimetric), ≥99.0% (HPLC), ≥98% (TLC)

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

assay

≥95.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

assay

≥97.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

Quality Level

200

Quality Level

400

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Fmoc-D-Asp(OtBu)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
852154000504027269091521
852154002504027269091538
852154100004027269212162
852154010004027269212155

Questions

Reviews

No rating value

Active Filters