Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 25 g | Check Cart for Availability | € 330,00 | |
| 100 g | Check Cart for Availability | € 986,00 |
About This Item
Quality Segment
product line
Novabiochem®
assay
≥97.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
mp
133-136 °C
application(s)
peptide synthesis
functional group
thiol
storage temp.
2-30°C
SMILES string
S(C(C)(C)C)CC(NC(=O)OCC1c2c(cccc2)c3c1cccc3)C(=O)O
InChI
1S/C22H25NO4S/c1-22(2,3)28-13-19(20(24)25)23-21(26)27-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,23,26)(H,24,25)
InChI key
IXAYZHCPEYTWHW-UHFFFAOYSA-N
General description
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Fmoc SPPS of Cysteine-Containing Peptides
Literature references
[1] S. N. McCurdy (1989) Pept. Res., 2, 147.
Analysis Note
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 97.0 %
Water (K. F.): ≤ 1.00 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form powder | form powder | form powder | form powder |
| assay ≥97.0% (acidimetric), ≥98.0% (HPLC), ≥98% (TLC) | assay ≥90.0% (acidimetric), ≥98% (TLC), ≥99.0% (HPLC) | assay ≥97.0% (HPLC), ≥97.0% (acidimetric), ≥98% (TLC) | assay ≥95.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC) |
| functional group thiol | functional group thiol | functional group thiol | functional group hydroxyl |
| product line Novabiochem® | product line Novabiochem® | product line Novabiochem® | product line Novabiochem® |
| storage temp. 2-30°C | storage temp. 15-25°C | storage temp. 2-30°C | storage temp. 15-25°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under Fmoc-Cys(tBu)-OH
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Overcome challenges in synthesis and disulfide bond formation with protocols for Fmoc solid-phase peptide synthesis of peptides with cysteine and methionine.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8520070100 | 04027269084745 |
| 8520070025 | 04027269084738 |



