Skip to Content
Merck
All Photos(1)

Key Documents

8.22298

Sigma-Aldrich

Phthalic acid

for synthesis

Synonym(s):

Phthalic acid, Benzene-1,2-dicarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€40.40
1 KG
€126.00
25 KG
€1,820.00

€40.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€40.40
1 KG
€126.00
25 KG
€1,820.00

About This Item

Linear Formula:
C6H4(COOH)2
CAS Number:
Molecular Weight:
166.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-873-2
NACRES:
NA.22

€40.40


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

7.8 hPa ( 191 °C)

Assay

≥99% (acidimetric)

form

solid

potency

7900 mg/kg LD50, oral (Rat)

pH

2 (20 °C, 5 g/L in H2O)

bp

289 °C/1013 hPa (decomposition)

mp

191 °C (closed capillary tube)

transition temp

flash point 168 °C

solubility

water: 5.74 g/L at 20 °C

density

1.59 g/cm3 at 20 °C

bulk density

960 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)c1c(cccc1)C(=O)[O-].[H+].[H+]

InChI

1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

XNGIFLGASWRNHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service