Skip to Content
Merck
All Photos(2)

Key Documents

125733

Sigma-Aldrich

Phthalic anhydride

ReagentPlus®, 99%

Synonym(s):

1,2-Benzenedicarboxylic anhydride, 1,3-Isobenzofurandione, Phthalic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4O3
CAS Number:
Molecular Weight:
148.12
Beilstein:
118515
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

5.1 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

flakes

autoignition temp.

1058 °F

expl. lim.

10.4 %

bp

284 °C (lit.)

mp

131-134 °C (lit.)

functional group

anhydride
ester

SMILES string

O=C1OC(=O)c2ccccc12

InChI

1S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H

InChI key

LGRFSURHDFAFJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phthalic anhydride is an industrially important chemical. It can be prepared from o-xylene or naphthalene.[1] It is widely used for the industrial preparation of plasticizers for PVC (polyvinyl chloride).[2]
Phthalic anhydride can be synthesized via oxidation of o-xylene, in the presence of V2O5/TiO2 monolithic catalyst supported by aluminium honeycomb.[3]

Application

Phthalic anhydride may be used in the synthesis of the following compounds:

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

305.6 °F - DIN 51758

Flash Point(C)

152 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Conductive monolithic catalysts: development and industrial pilot tests for the oxidation of o-xylene to phthalic anhydride"
Groppi G, et al.
Industrial & Engineering Chemistry Research, 51(22), 7590-7596 (2011)
Phthalic anhydride from o-xylene catalysis: science and engineering.
Nikolov V, et al.
Catalysis Reviews: Science and Engineering, 33(3-4), 319-374 (1991)
C J Sparham et al.
Journal of chromatography. A, 1062(1), 39-47 (2005-02-01)
A new method for the analysis of alcohol ethoxylates (AEs) using electrospray ionisation liquid chromatography/mass spectrometry (ESI LC/MS) is described. The procedure incorporates a novel derivatisation step with phthalic anhydride for the analysis of EO0-20 ethoxylates in a single analysis.
Yuichi Kajita et al.
Journal of the American Chemical Society, 130(51), 17226-17227 (2008-12-04)
An intermolecular nickel-catalyzed addition reaction has been developed where phthalic anhydrides react with alkynes to afford substituted isocoumarins. A mechanistic rationale is proposed, implying reductive elimination of Ni(0) promoted by ZnCl(2) cocatalyst as the key step of the catalytic cycle.
Xin Hai et al.
Electrophoresis, 31(19), 3352-3361 (2010-10-01)
A rapid and sensitive electrophoretically mediated microanalysis method with field-enhanced sample injection (FESI) for in-capillary derivatization was developed to determine selenomethionine (SeMet) and selenomethionine selenoxide (SeOMet). Phthalic anhydride (PA) was selected as the derivatization reagent due to the fast reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service