Skip to Content
Merck

Skip To

1.02401

Benzoic acid

volumetric standard, secondary reference material for alkalimetry, traceable to NIST Standard Reference Material (SRM) Certipur® Reag. Ph Eur,Reag. USP

Synonym(s):

Benzoic acid, Phenylformic acid, Benzene carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
60 g
Check Cart for Availability
€ 218,00
€ 185,30
60 g
Check Cart for Availability
€ 293,00
€ 219,75

About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
41116107
EC Index Number:
200-618-2
NACRES:
NA.24

€ 185,30

List Price€ 218,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ISO 17025, reag. Ph. Eur., reag. USP

vapor pressure

0.001 hPa ( 20 °C)

product line

Certipur®

form

solid

autoignition temp.

570 °C

potency

1700 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

technique(s)

titration: suitable

pH

2.5-3.5 (20 °C in H2O, saturated solution)

bp

249 °C/1013 hPa

mp

121-123 °C

transition temp

flash point 121 °C

solubility

2.9 g/L

density

1.321 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

15-25°C

SMILES string

[O-]C(=O)c1ccccc1.[H+]

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

General description

Benzoic acid is a volumetric standard produced under rigorous conditions and qualified with the highest possible precision. Its mass fraction is determined by potentiometric titration, and it is qualified in a DIN EN ISO/IEC 17025 accredited laboratory.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.


In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used as a certified reference material (CRM) for titer determinations in alkalimetric titrations, following USP and Ph. Eur.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • Certified reference material according to ISO 17034
  • High-purity material traceable to NIST SRMs
  • Measured in a DIN EN ISO/IEC 17025 accredited laboratory
  • In accordance with the chapter reagents of USP and Ph. Eur
  • High-quality offering accurate titer determinations

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
181021235333047
technique(s)

titration: suitable

technique(s)

-

technique(s)

titration: suitable

technique(s)

-

product line

Certipur®

product line

-

product line

-

product line

-

solubility

2.9 g/L

solubility

water: soluble (2.9 g/l at 25 °C)

solubility

water: soluble (2.9 g/l at 25 °C)

solubility

water: soluble (2.9 g/l at 25 °C)

potency

1700 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

potency

-

potency

-

potency

-

storage temp.

15-25°C

storage temp.

-

storage temp.

-

storage temp.

-

pH

2.5-3.5 (20 °C in H2O, saturated solution)

pH

3-5 (20 °C, 0.1%)

pH

-

pH

-


Still not finding the right product?

Explore all of our products under Benzoic acid


pictograms

CorrosionHealth hazard

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

The titer determination or standardization of a titration solution is essential for accurate and reliable titration results.





Global Trade Item Number

SKUGTIN
102401006004022536555775
102401006304054839567179

Questions

Reviews

No rating value

Active Filters